NECi Colorimetric Nitrate Water Test Kit, Low Range, 10/Pk
Representative image only.
NECi Colorimetric Nitrate Water Test Kit, Low Range, 10/Pk
| Mfr: NTK-WLR-10| Item: 0554325
Get quick, reliable non-toxic results
₹19,700.00INR / Pkg of 10
₹19,700.00INR / Pkg of 10
Product Description
Key Features
  • Eliminates the use of cadmium, a known carcinogen
  • Sensitive and accurate—no drift or inconsistency in testing results over time
  • Testing can be easily done in the field or laboratory
  • Enzyme-based nitrate test kits are environmentally friendly
  • 6 month warranty
More About this Item
Nitrate testing kits provide accurate and reliable results without the use of cadmium, a toxic heavy metal and known carcinogen. All kits use the naturally occurring enzyme called nitrate reductase which safely measures nitrate in any aqueous sample.
The laboratory nitrate analysis method has been validated and approved by ASTM International, USGS, and as an Alternate Test Procedure under the Safe Drinking Water Act with the US EPA.
6 Month Warranty
Specifications
  • Measurement Points0.05 - 2.0 ppm
  • DescriptionColorimetric Nitrate Water Test Kit, Low Range, 10/Pk