MTC™ Bio DuoClick™ Screw-Cap Plastic Culture Tubes
Dual position caps allow airflow for for aerobic culturing and airtight seal for anaerobic culturing
  • Flat, markable screw cap is easy to open and close
  • Two-click screw cap for aerobic and anaerobic culturing
The MTC™ Bio DuoClick™ culture tubes feature a unique screw closure system. DuoClick tubes eliminate the excess force, splashing, and general frustration of traditional snap-cap culture tubes. The advanced cap design includes two distinct positions for aerobic and anaerobic culturing. Just rotate the cap to the first click to secure the cap in the aerobic position or rotate it past the second click for a leakproof, airtight seal for anaerobic culturing.
The tubes are available in two plastic polymers, polystyrene (PS) and polypropylene (PP), and two capacities, 5- and 14-mL. Tubes are available sterile and non-sterile and in a variety of packaging configurations. Sterile tubes feature an attached cap and printed graduations, non-sterile tubes come bulk packed without caps attached and without graduations.
28,600.00 - ₹64,400.00INR / Case of 500
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sterile Tube Capacity (mL) Description
Availability Pricing
MTC Bio DuoClick™ Culture Tube with Screw Cap, Sterile, Polypropylene, 12 x 75 mm, 5 mL; 25/Bag, 500/Cs 3450100 Mfr # T8730
COMPARE
Yes 5 Culture Tube with Screw Cap, Sterile, Polypropylene, 12 x 75 mm, 5 mL; 25/Bag, 500/Cs
₹30,700.00INR / Case of 500
MTC Bio DuoClick™ Culture Tube with Screw Cap, Sterile, Polypropylene, 12 x 75 mm, 5 mL; 50/Rack, 500/Cs 3450101 Mfr # T8732
COMPARE
Yes 5 Culture Tube with Screw Cap, Sterile, Polypropylene, 12 x 75 mm, 5 mL; 50/Rack, 500/Cs
₹37,200.00INR / Case of 500
MTC Bio DuoClick™ Culture Tube with Screw Cap, Sterile, Polypropylene, 12 x 75 mm, 5 mL; 1/Bag, 500/Cs 3450102 Mfr # T8734
COMPARE
Yes 5 Culture Tube with Screw Cap, Sterile, Polypropylene, 12 x 75 mm, 5 mL; 1/Bag, 500/Cs
₹45,700.00INR / Case of 500
MTC Bio DuoClick™ Culture Tube with Screw Cap, Sterile, Polystyrene, 12 x 75 mm, 5 mL; 25/Bag, 500/Cs 3450103 Mfr # T8741
COMPARE
Yes 5 Culture Tube with Screw Cap, Sterile, Polystyrene, 12 x 75 mm, 5 mL; 25/Bag, 500/Cs
₹33,000.00INR / Case of 500
MTC Bio DuoClick™ Culture Tube with Screw Cap, Sterile, Polystyrene, 12 x 75 mm, 5 mL; 50/Rack, 500/Cs 3450104 Mfr # T8742
COMPARE
Yes 5 Culture Tube with Screw Cap, Sterile, Polystyrene, 12 x 75 mm, 5 mL; 50/Rack, 500/Cs
₹42,900.00INR / Case of 500
MTC Bio DuoClick™ Culture Tube with Screw Cap, Non-Sterile, Polypropylene, 12 x 75 mm, 5 mL; 1000/Cs 3450105 Mfr # T8750
COMPARE
No 5 Culture Tube with Screw Cap, Non-Sterile, Polypropylene, 12 x 75 mm, 5 mL; 1000/Cs
₹28,600.00INR / Case of 1000
MTC Bio DuoClick™ Culture Tube with Screw Cap, Sterile, Polypropylene, 17 x 100 mm, 14 mL; 25/Bag, 500/Cs 3450106 Mfr # T8830
COMPARE
Yes 14 Culture Tube with Screw Cap, Sterile, Polypropylene, 17 x 100 mm, 14 mL; 25/Bag, 500/Cs
₹45,700.00INR / Case of 500
MTC Bio Culture Tube, 14mL, 17 x 100mm, PP,  w/ attached 2-position screw-cap, printed graduations, sterile, 10 foam racks of 50 tubes, 500/CS 3450107 Mfr # T8832
COMPARE
Yes 14 Culture Tube with Screw Cap, Sterile, Polypropylene, 17 x 100 mm, 14 mL; 50/Rack, 500/Cs
₹56,100.00INR / Case of 500
MTC Bio DuoClick™ Culture Tube with Screw Cap, Sterile, Polypropylene, 17 x 100 mm, 14 mL; 1/Bag, 500/Cs 3450108 Mfr # T8834
COMPARE
Yes 14 Culture Tube with Screw Cap, Sterile, Polypropylene, 17 x 100 mm, 14 mL; 1/Bag, 500/Cs
₹64,400.00INR / Case of 500
MTC Bio DuoClick™ Culture Tube with Screw Cap, Sterile, Polystyrene, 17 x 100 mm, 14 mL; 25/Bag, 500/Cs 3450109 Mfr # T8840
COMPARE
Yes 14 Culture Tube with Screw Cap, Sterile, Polystyrene, 17 x 100 mm, 14 mL; 25/Bag, 500/Cs
₹50,800.00INR / Case of 500
MTC Bio DuoClick™ Culture Tube with Screw Cap, Sterile, Polystyrene, 17 x 100 mm, 14 mL; 50/Rack, 500/Cs 3450110 Mfr # T8842
COMPARE
Yes 14 Culture Tube with Screw Cap, Sterile, Polystyrene, 17 x 100 mm, 14 mL; 50/Rack, 500/Cs
₹60,200.00INR / Case of 500
MTC Bio DuoClick™ Culture Tube with Screw Cap, Non-Sterile, Polypropylene, 17 x 100 mm, 14 mL; 1000/Cs 3450111 Mfr # T8850
COMPARE
No 14 Culture Tube with Screw Cap, Sterile, Polypropylene, 17 x 100 mm, 14 mL; 1000/Cs
₹48,900.00INR / Case of 1000
Showing 1- 12 of 12