Monarch Instrument Track-It™ Barometric Pressure/Temperature Data Loggers
Monitor environmental conditions in critical areas
  • Record up to 64,000 nonvolatile barometric or temperature data points per logging session
  • Included software lets you easily program alarms, sampling rate, start/stop conditions, and units of measure
  • Direct-connect USB requires no additional cables
  • Altitude measurement: 0 to 30,000 ft (0 to 9144 m)
These barometric pressure (altitude) and temperature data loggers are ideal for monitoring cleanrooms and laboratories to make sure positive or negative pressure is maintained for safety. As a stand-alone data logger, the unit can record up to 64,000 data points before downloading to a PC. Onboard memory is nonvolatile, so data will not be lost if the battery dies. Alternatively, you can stream data in real time to a PC to keep an eye on changing conditions.
The multicolored LED indicator alerts you to alarm conditions and activity.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Monarch 5396-0321 Barometric Pressure & Temperature Datalogger w/ Display, standard battery WW-18011-12 Mfr # 5396-0321
COMPARE
Barometric Pressure & Temperature Datalogger w/ Display, standard battery
Call For Price
Monarch 5396-0322 Barometric Pressure & Temperature Datalogger w/ Display, long life battery WW-18011-13 Mfr # 5396-0322
COMPARE
Barometric Pressure & Temperature Datalogger w/ Display, long life battery