Monarch Instrument Pocket LED Stroboscopes
Designed to perform precision diagnostic inspections and rpm checks
  • Energy efficient with extremely bright, uniform light and long battery life
  • Quiet and cool operation
  • No lamp replacements
  • Built-in tripod mounting bushing (1/4 - 20)
  • Intuitive, one-handed operation
This pocket LED stroboscope is a compact, rugged, lightweight device that provides a super bright, uniform light output for performing visual diagnostic inspection and rpm measurements. The silent cool running LEDs are extremely energy efficient, providing up to five hours of operation on a single charge. Additional features include external input for remote triggering or tachometer mode, pulse output, memory for up to five preset flash rates, and tachometer mode for speed measurements up to 250,000 rpm using optional remote sensors and TTL pulse output.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Display Type Flash Duration Max RPM Range (rpm)
Availability Pricing
Monarch PLS Pocket LED Stroboscope, 115/230 VAC 5350001 Mfr # 6235-010
COMPARE
Backlit alphanumeric LCD 0.5 to 2500 µsec, 0.1 to 18°
Call For Price
Monarch PLS-Kit Pocket LED Stroboscope kit, 115/230 VAC 5350002 Mfr # 6235-011
COMPARE
Backlit alphanumeric LCD 0.5 to 2500 µsec, 0.1 to 18°