Millipore® Steriflip® Vacuum-Driven Filter Devices
For vacuum filtering solutions in 50-mL centrifuge tubes
  • Feature high-quality membranes
  • Millipore Express® (PES) membranes are ideal for low protein binding and exhibit faster flow rates with viscous solutions
  • Durapore® PVDF membranes offer the lowest protein binding available and are ideal for scale-up applications
  • Nylon mesh membranes are best for large particle removal
To filter, attach the unit directly to the 50-mL centrifuge tube containing your sample, flip it, and apply a vacuum. The filtrate collects directly in the attached 50-mL tube eliminating sample transfers between tubes. Disposable devices are sterile and have a 50-mL process volume.
Tube holder is not included.
59,000.00 - ₹112,000.00INR / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Membrane Pore Size (µm)
Availability Pricing
EMD Millipore SCGP00525 Steriflip-GP Filter, 0.22 um PES Express PLUS, Sterile; 25/Pk 2996920 Mfr # SCGP00525
COMPARE
Millipore Express® PES 0.22
₹112,000.00INR / Pkg of 25
EMD Millipore SE1M179M6 Steriflip-GP Filter, 0.22 um Durapore PVDF, Sterile; 25/Pk 2996924 Mfr # SE1M179M6
COMPARE
Durapore® PVDF 0.22
₹59,900.00INR / Pkg of 25
EMD Millipore SE1M003M00 Steriflip-GP Filter, 0.45 um Durapore PVDF, Sterile; 25/Pk 2996926 Mfr # SE1M003M00
COMPARE
Durapore® PVDF 0.45
₹59,000.00INR / Pkg of 25