EMD Millipore SE1M179M6 Steriflip-GP Filter, 0.22 um Durapore PVDF, Sterile; 25/Pk
Representative image only.
EMD Millipore SE1M179M6 Steriflip-GP Filter, 0.22 um Durapore PVDF, Sterile; 25/Pk
EMD Millipore | Mfr: SE1M179M6| Item: 2996924
For vacuum filtering solutions in 50-mL centrifuge tubes
₹59,900.00INR / Pkg of 25
₹59,900.00INR / Pkg of 25
Product Description
Key Features
  • Feature high-quality membranes
  • Millipore Express® (PES) membranes are ideal for low protein binding and exhibit faster flow rates with viscous solutions
  • Durapore® PVDF membranes offer the lowest protein binding available and are ideal for scale-up applications
  • Nylon mesh membranes are best for large particle removal
  • Durapore® PVDF membranes offer the lowest protein binding available and are ideal for scale-up applications
More About this Item
To filter, attach the unit directly to the 50-mL centrifuge tube containing your sample, flip it, and apply a vacuum. The filtrate collects directly in the attached 50-mL tube eliminating sample transfers between tubes. Disposable devices are sterile and have a 50-mL process volume.
Tube holder is not included.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Sterile
Specifications
  • Pore Size (µm)0.22
  • Tube Size (mL)50
  • MembraneDurapore® PVDF
  • Max Sample Volume (mL)50
  • DescriptionSteriflip-GP Filter, 0.22 um Durapore PVDF, Sterile; 25/Pk