Microplate/Microtube Shaking Incubator
Two interchangeable platforms for microplates and microtubes
  • Precise digital time, temperature and speed controls
  • Mechanical convection for uniform and stable heating
  • Cold room compatible for sub-ambient applications
What's included: Platforms for plates and tubes, aand a 6-ft power cord.
857,000.00 - ₹875,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Description
Availability Pricing
Labnet S2056A Microplate/Microtube Shaking Incubator - 120VAC 1731050 Mfr # S2056A
COMPARE
120 Microplate/Microtube Shaking Incubator - 120VAC
₹875,000.00INR / Each
Labnet S2056A-220V Microplate/Microtube Shaking Incubator; 220VAC 1731052 Mfr # S2056A-220
COMPARE
220 Microplate/Microtube Shaking Incubator - 220VAC
₹857,000.00INR / Each