Mettler Toledo XPR Essential Micro Balances
Offers fast, accurate micro weighing results
  • LevelControl warns when balance is not level
  • Draft shield dismantles completely for easy cleaning
  • Overload protection for load cell
  • User management and password protection
  • Multiple ports offer secure connectivity and data export
  • Color touch screen provides clear visibility — easy to use even with gloves on
  • StatusLight™ on terminal indicates when balance is ready for weighing
  • Fully Automatic Calibration Technology (FACT) performs internal calibrations automatically
  • SmartTrac program facilitates dosing in small increments to a target weight
These compact, front-loading XPR Essential micro balances allow you to accurately measure micro amounts in environments with limited lab bench space or fume hoods. The SmartPan weighing pan offers great stability, ensuring fast results. The 7-inch color touch-sensitive display provides easy accessibility, can be used with gloves on, and be positioned wherever it's most convenient in the lab.
The unique design of the balances protects the load cell and electronics from dust and spills, with smooth, rounded surfaces and a drip tray for easy cleaning. The one touch wing doors offer effortless access to the weighing chamber, giving you full access and visibility into the weighing chamber. The ergonomic design makes it easier to rest your arm while dosing, reducing strain during repetitive tasks. The integration with LabX software also adds a layer of automation, traceability, and data management.
The balances are also equipped with tolerance profiles to ensure weighing tasks meet defined quality requirements and regulations. In addition, the Good Weighing Practice® (GWP) Approved quality assurance function actively monitors the balance to confirm it is safe to weigh -- green light illuminates when the balance is ready to accept samples.
8,105,000.00 - ₹9,492,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity Dual Range Readability Dual Range Repeatability (mg)
Availability Pricing
Mettler Toledo XPR3DUE Micro Balance, 3.2/1.2 g x 0.01 mg, Internal Calibration, SmartTrac Dosing Guide 1100378 Mfr # 30900669
COMPARE
1.2/3.2 g 0.001/0.01 mg 0.001
₹8,105,000.00INR / Each
Mettler Toledo XPR16DUE Micro Balance, 12/3.2 g x 0.01 mg, Internal Calibration, SmartTrac Dosing Guide 1100379 Mfr # 30904616
COMPARE
3.2/12 g 0.001/0.01 mg 0.002
₹9,492,000.00INR / Each