Mettler Toledo MX Series NTEP-Approved Advanced Analytical Balances
High-resolution MonoBloc load cell delivers accurate and reliable results
  • NTEP-approved legal-for-trade balances meet government standards and requirements for commercial applications
  • 7" color TFT touchscreen display with intuitive user interface
  • Fully automatic time- and temperature-controlled internal calibration adjustment ensures accuracy
  • Unique SmartPan design leads to fast stabilization
  • Backlit draft shield for convenient handling and improved visibility
  • Flexible integration options—connect using Ethernet, USB-A, and USB-C
  • Automatic datalogging and user management for GxP guideline compliance
  • StatusLight provides added assurance that the prerequisites for valid results are met
  • Set testing schedule reminder to ensure your test results remain accurate
  • Durable high-grade chemical-resistant die-cast aluminum housing
The MX series NTEP-approved advanced analytical balances feature MonoBloc weighing technology and FACT (fully automatic time- and temperature-controlled internal adjustment) to provide accurate, repeatable results for everyday weighing tasks. The specially designed pan allows the balance to stabilize quickly even in turbulent environments. A high-contrast display with big numbers, clear symbols, and an intuitive menu with eight built-in applications allow users to operate the balance quickly and with ease.
Connect balance to a printer or computer using Ethernet or USB connection for complete and error-free documentation to easily comply with GxP guidelines. Draft shield doors open from either side for convenient sample handling. Remove, clean, and replace all draft shield panels in just a few simple steps, without tools; drip tray ensures quick cleaning of the balance.
A robust housing made of high-quality chemical-resistant aluminum protects the sensitive sensor from environmental influences and impacts. Leveling assistant feature indicates when the balance is not level and provides onscreen help to correct it. Password protection feature restricts modification of the balance settings to authorized personnel only. Overload protection guards the weighing cell against excess weight overload ensuring long service life.
2,153,000.00 - ₹3,361,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
Mettler Toledo MX105DU/A Analytical Balance, NTEP-Approved, MX Series, 42/120g x 0.01/0.1mg with Touchscreen, Backlit Draft Shield, and FACT Internal Calibration 0262844 Mfr # 30665092
COMPARE
120 0.1 Dual-Range Analytical Balance, NTEP-Approved, 42/120 g x 0.01/0.1 mg
₹2,758,000.00INR / Each
Mettler Toledo MX205DU/A Analytical Balance, NTEP-Approved, MX Series, 82/120g x 0.01/0.1mg with Touchscreen, Backlit Draft Shield, and FACT Internal Calibration 0262845 Mfr # 30665095
COMPARE
220 0.1 Dual-Range Analytical Balance, NTEP-Approved, 82/220 g x 0.01/0.1 mg
₹2,976,000.00INR / Each
Mettler Toledo MX105/A Analytical Balance, NTEP-Approved, MX Series, 120g x 0.01mg with Touchscreen, Backlit Draft Shield, and FACT Internal Calibration 0262846 Mfr # 30665138
COMPARE
120 0.01 Analytical Balance, NTEP-Approved, 120 g x 0.01 mg
₹3,361,000.00INR / Each
Mettler Toledo MX104/A Analytical Balance, NTEP-Approved, MX Series, 120g x 0.1mg with Touchscreen, Backlit Draft Shield, and FACT Internal Calibration 0262847 Mfr # 30665141
COMPARE
120 0.1 Analytical Balance, NTEP-Approved, 120 g x 0.1 mg
₹2,153,000.00INR / Each
Mettler Toledo MX204/A Analytical Balance, NTEP-Approved, MX Series, 220g x 0.1mg with Touchscreen, Backlit Draft Shield, and FACT Internal Calibration 0262848 Mfr # 30665144
COMPARE
220 0.1 Analytical Balance, NTEP-Approved, 220 g x 0.1 mg
₹2,392,000.00INR / Each
Mettler Toledo MX304/A Analytical Balance, NTEP-Approved, MX Series, 320g x 0.1mg with Touchscreen, Backlit Draft Shield, and FACT Internal Calibration 0262849 Mfr # 30665147
COMPARE
320 0.1 Analytical Balance, NTEP-Approved, 320 g x 0.1 mg
₹2,559,000.00INR / Each