Mettler Toledo MX Series Advanced NTEP-Approved Precision Toploading Balances
Fast settling time and consistent results for routine to complicated weighing procedures
  • NTEP-approved legal-for-trade balances meet government standards and requirements for commercial applications
  • High-resolution MonoBloc load cell delivers accurate and reliable results
  • Fully automatic time- and temperature-controlled internal calibration adjustment ensures accuracy
  • 7" color TFT touchscreen display with intuitive user interface
  • Unique SmartPan design leads to fast stabilization
  • Flexible integration options—connect using Ethernet, USB-A, and USB-C
  • Automatic datalogging and user management for GxP guideline compliance
  • StatusLight provides added assurance that the prerequisites for valid results are met
  • Set testing schedule reminder to ensure your test results remain accurate
  • Durable high-grade chemical-resistant die-cast aluminum housing
The MX series advanced NTEP-approved precision toploading balances feature MonoBloc weighing technology and FACT (fully automatic time- and temperature-controlled internal adjustment) to provide accurate, repeatable results for everyday weighing tasks. The specially designed pan allows the balance to stabilize quickly even in turbulent environments. A high-contrast display with big numbers, clear symbols, and an intuitive menu with nine built-in applications allow users to operate the balance quickly and with ease.
Connect balance to a printer or computer using Ethernet or USB connection for complete and error-free documentation to easily comply with GxP guidelines. Leveling assistant feature indicates when the balance is not level and provides onscreen help to correct it. A robust housing made of high-quality chemical-resistant aluminum protects the sensitive sensor from environmental influences and impacts. Password protection feature restricts modification of the balance settings to authorized personnel only. Overload protection guards the weighing cell against excess weight overload ensuring long service life.
1,377,000.00 - ₹3,136,000.00INR / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
Mettler Toledo MX303/A Precision Toploading Balance, NTEP-Approved, MX Series, 320g x 1mg with Touchscreen, Backlit Draft Shield, and FACT Internal Calibration 0262850 Mfr # 30665150
COMPARE
320 1 Precision Toploading Balance, NTEP-Approved, 320 g x 1 mg with Backlit Draft Shield
₹1,607,000.00INR / Each
Mettler Toledo MX303N/A Precision Toploading Balance, NTEP-Approved, MX Series, 320g x 1mg with Touchscreen and FACT Internal Calibration 0262851 Mfr # 30665153
COMPARE
320 1 Precision Toploading Balance, NTEP-Approved, 320 g x 1 mg
₹1,377,000.00INR / Each
Mettler Toledo MX603/A Precision Toploading Balance, NTEP-Approved, MX Series, 620g x 1mg with Touchscreen, Backlit Draft Shield, and FACT Internal Calibration 0262852 Mfr # 30665156
COMPARE
620 1 Precision Toploading Balance, NTEP-Approved, 620 g x 1 mg with Backlit Draft Shield
₹1,914,000.00INR / Each
Mettler Toledo MX603N/A Precision Toploading Balance, NTEP-Approved, MX Series, 620g x 1mg with Touchscreen and FACT Internal Calibration 0262853 Mfr # 30665159
COMPARE
620 1 Precision Toploading Balance, NTEP-Approved, 620 g x 1 mg
₹1,614,000.00INR / Each
Mettler Toledo MX1203/A Precision Toploading Balance, NTEP-Approved, MX Series, 1200g x 1mg with Touchscreen, Backlit Draft Shield, and FACT Internal Calibration 0262854 Mfr # 30665162
COMPARE
1200 1 Precision Toploading Balance, NTEP-Approved, 1200 g x 1 mg with Backlit Draft Shield
₹2,273,000.00INR / Each
Mettler Toledo MX1203N/A Precision Toploading Balance, NTEP-Approved, MX Series, 1200g x 1mg with Touchscreen and FACT Internal Calibration 0262855 Mfr # 30665165
COMPARE
1200 1 Precision Toploading Balance, NTEP-Approved, 1200 g x 1 mg
₹1,947,000.00INR / Each
Mettler Toledo MX2002/A Precision Toploading Balance, NTEP-Approved, MX Series, 2200g x 0.01g with Touchscreen and FACT Internal Calibration 0262856 Mfr # 30665168
COMPARE
2200 10 Precision Toploading Balance, NTEP-Approved, 2200 g x 0.01 g
₹1,540,000.00INR / Each
Mettler Toledo MX4002/A Precision Toploading Balance, NTEP-Approved, MX Series, 4200g x 0.01g with Touchscreen and FACT Internal Calibration 0262857 Mfr # 30665171
COMPARE
4200 10 Precision Toploading Balance, NTEP-Approved, 4200 g x 0.01 g
₹1,790,000.00INR / Each
Mettler Toledo MX6002/A Precision Toploading Balance, NTEP-Approved, MX Series, 6200g x 0.01g with Touchscreen and FACT Internal Calibration 0262858 Mfr # 30665174
COMPARE
6200 10 Precision Toploading Balance, NTEP-Approved, 6200 g x 0.01 g
₹1,962,000.00INR / Each
Mettler Toledo MX6002DR/A Precision Toploading Balance, NTEP-Approved, MX Series, 1200/6200g x 0.01/0.1g with Touchscreen and FACT Internal Calibration 0262859 Mfr # 30665177
COMPARE
6200 100 Dual-Range Precision Toploading Balance, NTEP-Approved, 1200/6200 g x 0.01/0.1 g
₹1,773,000.00INR / Each
Mettler Toledo MX12002/A Precision Toploading Balance, NTEP-Approved, MX Series, 12.2kg x 0.01g with Touchscreen and FACT Internal Calibration 0262860 Mfr # 30665180
COMPARE
12200 10 Precision Toploading Balance, NTEP-Approved, 12.2 kg x 0.01 g
₹2,392,000.00INR / Each
Mettler Toledo MX6001/A Precision Toploading Balance, NTEP-Approved, MX Series, 6200g x 0.1g with Touchscreen and FACT Internal Calibration 0262861 Mfr # 30665183
COMPARE
6200 100 Precision Toploading Balance, NTEP-Approved, 6200 g x 0.1 g
₹1,490,000.00INR / Each
Mettler Toledo MX8001/A Precision Toploading Balance, NTEP-Approved, MX Series, 8200g x 0.1g with Touchscreen and FACT Internal Calibration 0262862 Mfr # 30665186
COMPARE
8200 100 Precision Toploading Balance, NTEP-Approved, 8200 g x 0.1 g
₹1,683,000.00INR / Each
Mettler Toledo MX12001L/A Precision Toploading Balance, NTEP-Approved, MX Series, 12.2kg x 0.1g with Touchscreen and FACT Internal Calibration 0262863 Mfr # 30665189
COMPARE
12200 100 Precision Toploading Balance, NTEP-Approved, 12.2 kg x 0.1 g
₹2,342,000.00INR / Each
Mettler Toledo MX16001L/A Precision Toploading Balance, NTEP-Approved, MX Series, 16.2kg x 0.1g with Touchscreen and FACT Internal Calibration 0262864 Mfr # 30665192
COMPARE
16200 100 Precision Toploading Balance, NTEP-Approved, 16.2 kg x 0.1 g
₹2,647,000.00INR / Each
Mettler Toledo MX32001L/A Precision Toploading Balance, NTEP-Approved, MX Series, 32.2kg x 0.1g with Touchscreen and FACT Internal Calibration 0262865 Mfr # 30665195
COMPARE
32200 100 Precision Toploading Balance, NTEP-Approved, 32.2 kg x 0.1 g
₹3,136,000.00INR / Each
Mettler Toledo MX32000L/A Precision Toploading Balance, NTEP-Approved, MX Series, 32.2kg x 1g with Touchscreen and FACT Internal Calibration 0262866 Mfr # 30665198
COMPARE
32200 1000 Precision Toploading Balance, NTEP-Approved, 32.2 kg x 1 g
₹2,513,000.00INR / Each
Showing 1- 17 of 17