Mettler Toledo MR Series NTEP-Approved Analytical Balances
Easy worry-free weighing with accurate, reliable results day after day
  • NTEP-approved legal-for-trade balances meet government standards and requirements for commercial applications
  • Large color touchscreen and intuitive interface simplify operation
  • Electromagnetic Force Compensation (EMFC) weighing cell provides accurate and reliable results
  • Fully automatic time- and temperature-controlled internal calibration adjustment ensures accuracy
  • Flexible integration options—connect using Ethernet, USB-A, USB-C, and RS-232
  • Sample ID and passcode protection produce GLP documentation of results
  • Leveling assistant feature indicates when the balance is not level and offers onscreen help
  • Overload protection guards the weighing cell against excess weight overload
  • Remove, clean and replace all of the draft shield panels in just a few simple steps, without tools
  • Durable high-grade chemical-resistant die-cast aluminum housing
The MR series NTEP-approved analytical balances are great for your sensitive, yet routine weighing procedures. The bright 4.5” color TFT touchscreen with intuitive guidance enables you to operate quickly and efficiently, even when wearing gloves. An electromagnetic force compensation (EMFC) weighing cell with fully automated internal calibration technology (FACT) automatically adjusts the balance to compensate for a change in ambient temperature, to provide consistent reliable results.
Connect balance to a printer or computer using Ethernet, USB, or RS-232 connection for complete and error-free documentation to easily comply with GLP guidelines. Interfaces support a barcode reader, printer, PC, and other laboratory instruments for easy traceability and data handling. Eight embedded applications include essential weighing functions, statistics, and formulation. Restrict modification of the balance settings to authorized personnel only with the password protection feature.
A robust housing made of high-quality chemical-resistant aluminum protects the sensitive sensor from environmental influences and impacts. Smooth surfaces and rounded edges provide quick and easy cleaning. The QuickLock system keeps draft shield panels securely in place. Draft shield removes easily for cleaning; draft shield panels and drip tray are dishwasher safe. Built-in overload protection prolongs the life of your balance.
1,403,000.00 - ₹1,607,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (mg)
Availability Pricing
Mettler Toledo MR104/A Analytical Balance, NTEP-Approved, MR Series, 120g x 0.1mg with Touchscreen, Draft Shield, and FACT Internal Calibration 0262888 Mfr # 30666204
COMPARE
Internal 120 0.1
₹1,403,000.00INR / Each
Mettler Toledo MR204/A Analytical Balance, NTEP-Approved, MR Series, 220g x 0.1mg with Touchscreen, Draft Shield, and FACT Internal Calibration 0262889 Mfr # 30666207
COMPARE
Internal 220 0.1
₹1,506,000.00INR / Each
Mettler Toledo MR304/A Analytical Balance, NTEP-Approved, MR Series, 320g x 0.1mg with Touchscreen, Draft Shield, and FACT Internal Calibration 0262890 Mfr # 30666210
COMPARE
Internal 320 0.1
₹1,607,000.00INR / Each