Mettler Toledo Excellence XPR Precision Toploading Balances
Built-in quality assurance monitoring ensures data integrity
  • StatusLight™ uses colored lights on terminal to indicate when balance is ready for weighing
  • Performs temperature-activated internal calibrations automatically at preprogrammed intervals
  • Integrated LabX® software automatically saves all data results and provides high process security
  • SmartPan™ weighing pan reduces effect of turbulence, shortening stabilization time and repeatability
  • Graphical leveling device provides warning when balance is not level
  • Overload protection safeguards weighing cell against damage
  • Built-in notepad automatically records all task parameters and results
  • 7-inch color touch screen provides clear visibility of weighing results—is glove compatible
  • Password-protected individual profiles
The XPR series balances feature the GWP approved built-in quality assurance functions which automatically monitor the balance level, routine tests, and minimum weight limit, ensuring your results are always valid and comply with the strictest regulations. State-of-the-art user interface with intelligent functions helps simplify and supports your processes at every step.
The innovative SmartPan weighing pan minimizes air currents on the weighing cell, improving stabilization time and repeatability, allowing you to get results twice as fast as standard weighing pans without the use of a draft shield.
An intuitive touch screen streamlines operations, provides quick access to menus and tasks, and allows you to set up shortcuts and use multiple fingers to enter data. Store regularly used task methods in the method library for quick future recall. All task parameters are recorded on the results notepad where you can add your own comments. Easily transfer data to a PC, printer, or bar code reader via four USB or Ethernet ports. No more manual transcriptions.
The balances are designed with chemical-resistant materials, smooth surfaces, and rounded edges, making them easy to clean.
3,304,000.00 - ₹5,125,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (mg)
Availability Pricing
Mettler Toledo XPR12001L Excellence XPR Precision Toploader Balance, Large, 12.1kg x, 0.1g, Touchscreen 1133800 Mfr # 30875596
COMPARE
Internal 12100 100
Call For Price
Mettler Toledo XPR32001L Excellence XPR Precisions Toploader Balance, Large, 32.1kg x, 0.1g, Touchscreen 1133804 Mfr # 30316292
COMPARE
Internal 32100 100
Call For Price
Mettler Toledo XPR64000L Excellence XPR Precisions Toploader Balance, Large, 64.1kg x, 1g, Touchscreen 1133810 Mfr # 30316493
COMPARE
Internal 64100 1000
Call For Price
Mettler Toledo XPR603S Excellence Precision XPR Toploader Balance, 610g x, 1mg, SmartPan Weighing Pan, Touchscreen 1133828 Mfr # 30316504
COMPARE
Internal 610 1
Call For Price
Mettler Toledo XPR4002S Excellence Precision XPR Toploader Balance, 4.1kg x, 0.01g, SmartPan Weighing Pan, Touchscreen 1133850 Mfr # 30317269
COMPARE
Internal 4100 10
Call For Price
Mettler Toledo XPR10002S Excellence XPR Precisions Toploader Balance, 10.1kg x, 0.01g, SmartPan Weighing Pan, Touchscreen 1133858 Mfr # 30317433
COMPARE
Internal 10100 10
Call For Price
Mettler Toledo XPR1203SN Excellence XPR Precision Toploader Balance, Large , 1201g x , 0.001g , Touchscreen 1134895 Mfr # 30710392
COMPARE
Internal or External 1210 1
₹4,350,000.00INR / Each
Mettler Toledo XPR2003S Excellence XPR Precision Toploader Balance, Large , 2100g x , 0.001g , Touchscreen 1134897 Mfr # 30710397
COMPARE
Internal or External 2100 1
₹4,787,000.00INR / Each
Mettler Toledo XPR3004SD5 Excellence XPR Precision Toploader Balance, Large , 3100g x , 0.0005g , Touchscreen 1134899 Mfr # 30710393
COMPARE
Internal or External 3100 0.5
₹5,125,000.00INR / Each
Mettler Toledo XPR12002S Excellence XPR Precision Toploader Balance, Large , 12100g x , 0.01g , Touchscreen 1134900 Mfr # 30710395
COMPARE
Internal or External 12100 10
₹4,647,000.00INR / Each
Mettler Toledo XPR12003SD5 Excellence XPR Precision Toploader Balance, Large , 12100g x , 0.0005g , Touchscreen 1134902 Mfr # 30710394
COMPARE
Internal or External 12100 5
₹4,582,000.00INR / Each
Mettler Toledo XPR12001S Excellence XPR Precision Toploader Balance, Large , 12100g x , 0.1g , Touchscreen 1134903 Mfr # 30710396
COMPARE
Internal or External 12100 100
₹3,304,000.00INR / Each
Showing 1- 12 of 12