META Unisex Twill Lab Coats

Fabric features soil release properties for easier washing
Coats are made of 65% polyester, 35% cotton, fine line twill with a pleated back and sewn down belt for a tailored look. Lab coats are generously cut with double-sewn cuffs to extend sleeve lengths as necessary. Lab coats are equipped with two lower pockets, one chest pocket with pen divider, and two interior pockets — carry objects where it is most comfortable for you. Side slits allow access to clothing underneath lab coat. Unisex.
6,600.00 - ₹7,200.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Size
Availability Pricing
Meta 1963-36-011-S Unisex Twill Lab Coat, White, 40" L, Small WW-86468-20 Mfr # 1963-011-36
COMPARE
White 38 Small
₹7,200.00INR / Each
Meta 1963-38-011-M Unisex Twill Lab Coat, White, 40" L, Medium WW-86468-22 Mfr # 1963-011-38
COMPARE
White 38 Medium
₹7,200.00INR / Each
Meta 1963-42-011-L Unisex Twill Lab Coat, White, 40" L, Large WW-86468-24 Mfr # 1963-011-42
COMPARE
White 38 Large
₹6,600.00INR / Each
Meta 1963-48-011- Unisex Twill Lab Coat, White, 40" L, X-Large WW-86468-26 Mfr # 1963-011-48
COMPARE
White 38 X-Large
₹6,600.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat