Memmert Pump Module for VO Vacuum Ovens
Saves up to 70% in energy
  • Comes complete with cabinet to stack with your Memmert VO Vacuum oven (sold separately)
  • Gas-tight and maintenance-free
The noise-insulated pump modules used in combination with the VO vacuum ovens, feature a built-in, chemical-resistant vacuum pump. This variable-speed pump provides an energy savings of up to 70% in ramp mode and longer service life of membranes. Automatically detected by the VO vacuum ovens, the self-drying pump makes it possible during evacuation to blow the condensed liquid out of the pump heads at high speed, while maintaining the vacuum in the oven at a constant level.
The pump module easily stacks with your Memmert vacuum oven.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free-Air Capacity (LPM) Max Vacuum (mbar) Description
Availability Pricing
Memmert PMP 49 Variable-Speed Pump Module for VO49 Vacuum Oven; 230 VAC 5220121 Mfr # PMP 49
COMPARE
55 2 Variable-Speed Pump Module for VO49; 230 VAC
Call For Price
Memmert PMP 101 Variable-Speed Pump Module for VO101 Vacuum Oven; 230 VAC 5220122 Mfr # PMP 101
COMPARE
55 2 Variable-Speed Pump Module for VO101 Vacuum Oven; 230 VAC