Megger MPAC128-Series Acoustic Imaging Cameras
Locate gas leaks and partial discharges to increase safety and minimize costly equipment failures
  • 128 microphone array provides excellent sound detection
  • Detects leaks and discharges from up to 120 meters away
  • Effective test bandwidth from 2 to 48 kHz
  • 8 MP camera offers clear images
  • Simple operation with only two parameters to set
  • 8 GB storage capacity
  • Focus function reduces unwanted environmental noises
  • Phase Resolved Partial Discharge (PRPD) helps differentiate between different types of partial discharge
  • Converts ultrasonic sounds to audible sounds
  • Supports multiple languages
  • IP54-rated for protection against water and dust
The MPAC128 imaging cameras feature 128 microphones to capture sound information to quickly and accurately locate gas leaks and partial discharge threats. With the ability to detect quiet noises from up to 120 m (394 ft) away, these cameras provide efficient troubleshooting, prevent safety hazards, and minimize production disruptions, saving valuable time, energy, and resources.
Use the imaging cameras to easily detect multiple gas types and pressurized gas leaks. The cameras use microphone array beamforming technology to acquire and measure the sound source, then convert the sound to a visual image. View the captured sound transposed to an 8 MP digital camera displayed on a 7” color touch screen display.
Operating the camera is simple with only two parameters to set—the measurement frequency range and the dynamic range. They offer both picture and video capture modes, making data recording easy and flexible. Download test results to a removable microSD card or a USB-C flash drive for analysis and report generation. Multi-language feature offers operation in English, Dutch, French, Chinese, German, Hungarian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese, Russian, Spanish, or Swedish.
The hand straps and shoulder strap allow easy transportation and operation; mount the camera on a tripod using a tripod mount thread for hands-free operation. The durable camera comes with an IP54-rated housing for protection against water and dust. Use it in temperatures ranging from –20 to 50 °C (–4 to 122 °F).
5,780,000.00 - ₹10,789,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Field Of View (FOV) Pixels / Thermal Resolution Description
Availability Pricing
Megger MPAC128 Acoustic Imaging Camera 2002740 Mfr # MPAC128
COMPARE
Horiontal: 62°<br>Vertical: 48° 8 megapixels Acoustic Imaging Camera
₹5,780,000.00INR / Each
Megger MPAC128-ATEX Acoustic Imaging Camera 2002741 Mfr # MPAC128-ATEX
COMPARE
Horiontal: 62°<br>Vertical: 48° 8 megapixels ATEX-Certified Acoustic Imaging Camera
₹10,789,000.00INR / Each