Megger MPAC128-ATEX Acoustic Imaging Camera

Representative image only.

Megger MPAC128-ATEX Acoustic Imaging Camera

Megger | Mfr: MPAC128-ATEX| Item: 2002741

Locate gas leaks and partial discharges to increase safety and minimize costly equipment failures

₹10,789,000.00INR / Each

₹10,789,000.00INR / Each
Product Description

Includes

Hand strap, shoulder strap, headphone, cleaning tool, USB-A microSD card reader, 8 GB microSD card, 8 GB USB-C/USB-A memory stick, USB-C charging cable, universal charger, and protective carrying case.

Key Features

  • 128 microphone array provides excellent sound detection
  • Detects leaks and discharges from up to 120 meters away
  • Effective test bandwidth from 2 to 48 kHz
  • 8 MP camera offers clear images
  • Simple operation with only two parameters to set
  • 8 GB storage capacity
  • Focus function reduces unwanted environmental noises
  • Phase Resolved Partial Discharge (PRPD) helps differentiate between different types of partial discharge
  • Converts ultrasonic sounds to audible sounds
  • Supports multiple languages
  • IP54-rated for protection against water and dust
  • ATEX certified for use in hazardous Zone 2 areas
  • 2 year warranty

More About this Item

The MPAC128 imaging cameras feature 128 microphones to capture sound information to quickly and accurately locate gas leaks and partial discharge threats. With the ability to detect quiet noises from up to 120 m (394 ft) away, these cameras provide efficient troubleshooting, prevent safety hazards, and minimize production disruptions, saving valuable time, energy, and resources.

Use the imaging cameras to easily detect multiple gas types and pressurized gas leaks. The cameras use microphone array beamforming technology to acquire and measure the sound source, then convert the sound to a visual image. View the captured sound transposed to an 8 MP digital camera displayed on a 7” color touch screen display.

Operating the camera is simple with only two parameters to set—the measurement frequency range and the dynamic range. They offer both picture and video capture modes, making data recording easy and flexible. Download test results to a removable microSD card or a USB-C flash drive for analysis and report generation. Multi-language feature offers operation in English, Dutch, French, Chinese, German, Hungarian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese, Russian, Spanish, or Swedish.

The hand straps and shoulder strap allow easy transportation and operation; mount the camera on a tripod using a tripod mount thread for hands-free operation. The durable camera comes with an IP54-rated housing for protection against water and dust. Use it in temperatures ranging from –20 to 50 °C (–4 to 122 °F).

The MAPC128-ATEX acoustic imaging camera comes with a II 3G Ex ic IIC T5 Gc explosion-proof rating. It is specially designed for use in Zone 2 hazardous areas that may contain flammable gases, vapor, or mist in the atmosphere.

Specifications
  • Pixels / Thermal Resolution8 megapixels
  • Field Of View (FOV)Horiontal: 62°
    Vertical: 48°
  • Palette SelectionGrayscale, ironbow, or rainbow
  • Text AnnotationYes
  • Video OutputMicroSD card or USB-C memory stick
  • Data OutputMicroSD card or USB-C memory stick
  • Memory Storage TypePicture and video
  • Memory Capacity8 GB
  • Software IncludedYes
  • Display TypeCapacitive touch screen
  • Low Battery IndicatorYes
  • Battery lifeApproximately 4 hours (fully charged)
  • BatteryRechargeable Li-ion
  • Power (VAC)100 to 240
  • Height (in)6 7/8
  • Width (in)10 3/4
  • Depth (in)1.6875
  • DescriptionATEX-Certified Acoustic Imaging Camera
  • Warranty2 years
Reviews
Q&A