Marsh Bellofram Type 1500 I/P Transducers
Install in limited-space locations or manifold
  • 4 to 20 mA Input
  • Small footprint, compact size
  • Built-in volume booster
  • External zero and span adjustments
Transducers are an electro-pneumatic device that reduce supply pressure to a regulated output pressure directly proportional to an electrical input signal. They provide precise control to actuators, valves, and positioners, and are ideally used for high-flow devices. Supply and output ports are located on front and back of units, enabling simple installation and connections. Choose from a general-purpose or NEMA 4X enclosure.
56,600.00 - ₹143,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Control Range (high) (PSI) Forward Flow Capacity (LPM) Description
Availability Pricing
Marsh Bellofram 969-716-000 : 4-20MA-0-120PSI 6845834 Mfr # 969-716-000
COMPARE
₹143,000.00INR / Each
Marsh Bellofram 010-606-001:5 ST. MANIFOLD 6845840 Mfr # 010-606-001
COMPARE
₹56,600.00INR / Each
Marsh Bellofram 010-606-002:10 ST.MANIFOLD 6845841 Mfr # 010-606-002
COMPARE
₹81,100.00INR / Each
Marsh Bellofram 969-710-001 Pressure Transmitter, I to P;3-15 PSI, 4-20 mA; 1/2" Con 6882606 Mfr # 969-710-001
COMPARE
15 677 Pressure Transmitter, I to P;3-15 PSI, 4-20 mA; 1/2" Con
₹100,000.00INR / Each
Marsh Bellofram 969-710-101 Pressure Transmitter, I to P;3-15 PSI, 4-20 mA; Term Blk 6882612 Mfr # 969-710-101
COMPARE
15 677 Pressure Transmitter, I to P;3-15 PSI, 4-20 mA; Term Blk
₹100,000.00INR / Each
Marsh Bellofram 969-712-101 Pressure Transmitter, I to P;6-30 PSI, 4-20 mA; Term Blk 6882616 Mfr # 969-712-101
COMPARE
30 677 Pressure Transmitter, I to P;6-30 PSI, 4-20 mA; Term Blk
₹100,000.00INR / Each
Marsh Bellofram 969-710-000 Pressure Transmitter, I to P;3-15 PSI, 4-20 mA; NEMA4X 6882618 Mfr # 969-710-000
COMPARE
15 677 Pressure Transmitter, I to P;3-15 PSI, 4-20 mA; NEMA4X
₹120,000.00INR / Each
Showing 1- 7 of 7