Marsh Bellofram 010-606-001:5 ST. MANIFOLD
Representative image only.
Marsh Bellofram 010-606-001:5 ST. MANIFOLD
Marsh Bellofram | Mfr: 010-606-001| Item: 6845840
Install in limited-space locations or manifold
₹56,600.00INR / Each
₹56,600.00INR / Each
Product Description
Key Features
  • 4 to 20 mA Input
  • Small footprint, compact size
  • Built-in volume booster
  • External zero and span adjustments
  • 18 month warranty
More About this Item
Transducers are an electro-pneumatic device that reduce supply pressure to a regulated output pressure directly proportional to an electrical input signal. They provide precise control to actuators, valves, and positioners, and are ideally used for high-flow devices. Supply and output ports are located on front and back of units, enabling simple installation and connections. Choose from a general-purpose or NEMA 4X enclosure.
18 Month Warranty
Specifications
  • Warranty18 months