Marsh Bellofram Type 10 Air Pressure Regulators
Provide precision control in adverse conditions
  • Use with clean, dry air
  • Ideal for gas mixing, valve operations, and calibration stands
These pressure regulators are designed for precise regulation of air pressure in pipes, vessels, and process lines. Regulators maintain constant pressure by providing high accuracy, precision control, and maximum stability under the most adverse conditions. Regulators feature high-accuracy pressure regulation up to ±0.1%, a servo-operated control mechanism for pressure stability, and start-up stability that returns to output setting after a long down-time. Locking nut prevents inadvertant adjustment of pressure. Unit can be in-line or panel mounted.
26,000.00 - ₹31,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Max Exhaust Flow Rate (SCFM) Max Supply Pressure (PSI) Supply Pressure Effect
Availability Pricing
Marsh Bellofram 960-001-000 Pressure Regulator, 1/8" NPT(F), 2-25 psi 6882500 Mfr # 960-001-000
COMPARE
150 0.005 psi per 25 psi change
₹31,000.00INR / Each
Marsh Bellofram 960-007-000 Pressure Regulator, 1/8" NPT(F), 2-60 psi 6882502 Mfr # 960-007-000
COMPARE
150 0.005 psi per 25 psi change
₹26,000.00INR / Each
Marsh Bellofram 960-013-000 Pressure Regulator, 1/8" NPT(F), 2-120 psi 6882504 Mfr # 960-013-000
COMPARE
150 0.005 psi per 25 psi change
₹31,000.00INR / Each
Marsh Bellofram 960-003-000 Pressure Regulator, 1/4" NPT(F), 2-25 psi 6882506 Mfr # 960-003-000
COMPARE
150 0.005 psi per 25 psi change
₹31,000.00INR / Each
Marsh Bellofram 960-009-000 Pressure Regulator, 1/4" NPT(F), 2-60 psi 6882508 Mfr # 960-009-000
COMPARE
150 0.005 psi per 25 psi change
₹31,000.00INR / Each
Marsh Bellofram 960-015-000 Pressure Regulator, 1/4" NPT(F), 2-120 psi 6882510 Mfr # 960-015-000
COMPARE
150 0.005 psi per 25 psi change
₹31,000.00INR / Each
Bellofram 960-005-000 Pressure Regulator, 3/8" NPT(F), 2-25 psi 6882512 Mfr # 960-005-000
COMPARE
150 0.005 psi per 25 psi change
₹31,000.00INR / Each
Marsh Bellofram 960-011-000 Pressure Regulator, 3/8" NPT(F), 2-60 psi 6882514 Mfr # 960-011-000
COMPARE
150 0.005 psi per 25 psi change
₹31,000.00INR / Each
Marsh Bellofram 960-017-000 Pressure Regulator, 3/8" NPT(F), 2-120 psi 6882516 Mfr # 960-017-000
COMPARE
150 0.005 psi per 25 psi change
₹31,000.00INR / Each
Showing 1- 9 of 9