Mark-10 R51 Universal Torque Sensors with Interchangeable Grips

Outstanding overload protection
  • Interchangeable chucks (order separately)
  • Warranty: 3 years
A great solution for general bi-directional torque testing applications requiring different sample sizes. Three interchangeable chuck capacities and bit holder are available separately.
249,000.00 - ₹281,000.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (oz-in) Description
Availability Pricing
Mark-10 MR51-50Z Torque Sensor, 50 ozFin / 3600 gFcm / 36 kgFmm / 350 Nmm / 35 Ncm 2530232 Mfr # MR51-50Z
COMPARE
50 Plug & Test Universal Torque Sensor, Interchange Grips; 35 N cm
₹281,000.00INR / Each
Mark-10 MR51-12 Torque Sensor, 12 lbFin / 1 lbFft / 192 ozFin / 135 Ncm / 1.35 Nm 2530233 Mfr # MR51-12
COMPARE
192 Plug & Test Universal Torque Sensor, Interchange Grips; 135 N cm
₹261,000.00INR / Each
Mark-10 MR51-50 Torque Sensor, 50 lbFin / 4 lbFft / 800 ozFin / 570 Ncm / 5.7 Nm 2530234 Mfr # MR51-50
COMPARE
800 Plug & Test Universal Torque Sensor, Interchange Grips; 570 N cm
₹249,000.00INR / Each
Mark-10 MR51-100 Torque Sensor, 100 lbFin / 8 lbFft / 1600 ozFin / 1150 Ncm / 11.5 Nm 2530235 Mfr # MR51-100
COMPARE
1600 Plug & Test Universal Torque Sensor, Interchange Grips; 1150 N cm
₹261,000.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat