Mark-10 R02 Series Compact Compression Force Sensors
Small size
  • Stainless steel construction
  • Weight: 0.4 lb (0.2 kg)
  • Warranty: 3 years
Compact and rugged, the R02 is ideal for heavy duty applications with space constraints for compressive load testing.
249,000.00 - ₹284,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs)
Availability Pricing
Mark-10 MR02-100 Force Sensor, 100 lbF / 1600 ozF / 50 kgF / 50000 gF / 500 N 2530215 Mfr # MR02-100
COMPARE
50 100
₹249,000.00INR / Each
Mark-10 MR02-200 Force Sensor, 200 lbF / 3200 ozF / 100 kgF / 1000 N / 1 kN 2530216 Mfr # MR02-200
COMPARE
100 200
₹249,000.00INR / Each
Mark-10 MR02-500 Force Sensor, 500 lbF / 8000 ozF / 250 kgF / 2500 N / 2.5 kN 2530217 Mfr # MR02-500
COMPARE
250 500
₹249,000.00INR / Each
Mark-10 MR02-1000 Force Sensor, 1000 lbF / 16000 ozF / 500 kgF / 5000 N / 5 kN 2530218 Mfr # MR02-1000
COMPARE
500 1000
₹249,000.00INR / Each
Mark-10 MR02-2000 Force Sensor, 2000 lbF / 32000 ozF / 1000 kgF / 10000 N / 10 kN 2530219 Mfr # MR02-2000
COMPARE
1000 2000
₹249,000.00INR / Each
Mark-10 MR02-5000 Force Sensor, 5000 lbF / 2500 kgF / 25000 N / 25 kN 2530220 Mfr # MR02-5000
COMPARE
2500 5000
₹284,000.00INR / Each
Mark-10 MR02-10000 Force Sensor, 10000 lbF / 5000 kgF / 50000 N / 50 kN 2530221 Mfr # MR02-10000
COMPARE
5000 10000
₹284,000.00INR / Each
Showing 1- 7 of 7