Mark-10 Plug & Test Force/Torque Indicators
Test up to 10,000 lb of force and 5000 lb in. of torque with interchangeable sensors
These force/torque indicators are compatible with Mark-10’s Plug & Test™ family of interchangeable force and torque sensors. All calibration and configuration data is saved within the sensor’s smart connector, not the indicator, allowing for true interchangeability. The included software allows you to further analyze measurement results and is ideal for report generation requirements. Probe is needed for calibration.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Sampling Rate Description
Availability Pricing
Mark-10 M3i Basic Force/Torque Indicator 2530176 Mfr # M3I
COMPARE
2000 Hz Basic Force/Torque Gauge
Call For Price
Mark-10 M7i Professional Force/Torque Indicator 2530177 Mfr # M7I
COMPARE
14,000 Hz Professional Force/Torque Gauge
Call For Price
Mark-10 M5i Advanced Force/Torque Indicator 2530200 Mfr # M5I
COMPARE
7000 Hz Advanced Force/Torque Gauge