Low-Pressure Tyvek-Hood Supplied-Air Respirator Kits
Complete and convenient NIOSH-approved systems you can take anywhere
  • Large window provides maximum view
  • Ideal for worker with facial hair
Complete, continuous-flow low-pressure respirator systems for one or two workers are NIOSH approved. These convenient systems come in one box with one part number for easy ordering. Each system has respirator(s), breathing tube(s), air hose(s) and ambient air pump.
Tyvek® Hood systems are excellent for workers with facial hair who can not wear tight-fitting face pieces. The lightweight disposable hoods feature a large window for maximum view. Hood systems are ideal for applications such as painting, or asbestos abatement.
Approval: NIOSH (as a system)
694,000.00 - ₹1,588,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Allegro 9220-01 One Man Tyvek Hood Supplied Air System with 50 foot Hose 8152013 Mfr # 9220-01
COMPARE
One Man Tyvek Hood Supplied Air System with 50 foot Hose
₹694,000.00INR / Each
Allegro 9220-02 Two Man Tyvek Hood Supplied Air System with 50 foot Hose 8152017 Mfr # 9220-02
COMPARE
Two Man Tyvek Hood Supplied Air System with 50 foot Hose
₹1,446,000.00INR / Each
Allegro 9230-02 Two Man Tyvek Hood Supplied Air System with 100 Foot Hose 8152019 Mfr # 9230-02
COMPARE
Two Man Tyvek Hood Supplied Air System with 100 Foot Hose
₹1,588,000.00INR / Each