Allegro 9220-02 Two Man Tyvek Hood Supplied Air System with 50 foot Hose
Representative image only.
Allegro 9220-02 Two Man Tyvek Hood Supplied Air System with 50 foot Hose
Allegro | Mfr: 9220-02| Item: 8152017
Complete and convenient NIOSH-approved systems you can take anywhere
₹1,446,000.00INR / Each
₹1,446,000.00INR / Each
Buy It Together
Current Item
Allegro 9220-02 Two Man Tyvek Hood Supplied Air System with 50 foot Hose
Allegro 9220-02 Two Man Tyvek Hood Supplied Air System with 50 foot Hose
8152017
₹1,446,000.00INR / Each
+ =
Total Price 1478000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Large window provides maximum view
  • Ideal for worker with facial hair
  • 1 year warranty
More About this Item
Complete, continuous-flow low-pressure respirator systems for one or two workers are NIOSH approved. These convenient systems come in one box with one part number for easy ordering. Each system has respirator(s), breathing tube(s), air hose(s) and ambient air pump.
Tyvek® Hood systems are excellent for workers with facial hair who can not wear tight-fitting face pieces. The lightweight disposable hoods feature a large window for maximum view. Hood systems are ideal for applications such as painting, or asbestos abatement.
Approval: NIOSH (as a system)
1 Year Warranty
Specifications
  • ComplianceNIOSH
  • DescriptionTwo Man Tyvek Hood Supplied Air System with 50 foot Hose