Low-Pressure Full-Facepiece Supplied-Air Respirator Kits
Complete and convenient NIOSH-approved systems you can take any where
Complete, continuous-flow low-pressure respirator systems for one or two workers are NIOSH approved. These convenient kits come in one box with one part number for easy ordering. Each system has respirator(s), breathing tube(s), air hose(s) and ambient air pump.
Full-face-piece systems feature soft silicone masks with double-sealing flange for improved mask-to-face seal. Mask also features a removable hair net and fits under a hard hat. Replacement face piece available.
Approval: NIOSH (as a system)
645,000.00 - ₹1,107,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Allegro 9200-01 One Man Full Face Piece Supplied Air System with 50 foot Hose 8152003 Mfr # 9200-01
COMPARE
One Man Full Face Piece Supplied Air System with 50 foot Hose
₹645,000.00INR / Each
Allegro 9210-01 One Man Full Face Piece Supplied Air System with 100 foot Hose 8152005 Mfr # 9210-01
COMPARE
One Man Full Face Piece Supplied Air System with 100 foot Hose
₹687,000.00INR / Each
Allegro 9200-02 Two Man Full Face Piece Supplied Air System with 50 foot Hose 8152007 Mfr # 9200-02
COMPARE
Two Man Full Face Piece Supplied Air System with 50 foot Hose
₹1,006,000.00INR / Each
Allegro 9210-02 Two Man Full Face Piece Supplied Air System with 100 foot Hose 8152009 Mfr # 9210-02
COMPARE
Two Man Full Face Piece Supplied Air System with 100 foot Hose
₹1,107,000.00INR / Each