Allegro 9210-01 One Man Full Face Piece Supplied Air System with 100 foot Hose
Representative image only.
Allegro 9210-01 One Man Full Face Piece Supplied Air System with 100 foot Hose
Allegro | Mfr: 9210-01| Item: 8152005
Complete and convenient NIOSH-approved systems you can take any where
₹687,000.00INR / Each
₹687,000.00INR / Each
Buy It Together
Current Item
Allegro 9210-01 One Man Full Face Piece Supplied Air System with 100 foot Hose
Allegro 9210-01 One Man Full Face Piece Supplied Air System with 100 foot Hose
8152005
₹687,000.00INR / Each
+ =
Total Price 829000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
Complete, continuous-flow low-pressure respirator systems for one or two workers are NIOSH approved. These convenient kits come in one box with one part number for easy ordering. Each system has respirator(s), breathing tube(s), air hose(s) and ambient air pump.
Full-face-piece systems feature soft silicone masks with double-sealing flange for improved mask-to-face seal. Mask also features a removable hair net and fits under a hard hat. Replacement face piece available.
Approval: NIOSH (as a system)
1 Year Warranty
Specifications
  • ComplianceNIOSH
  • DescriptionOne Man Full Face Piece Supplied Air System with 100 foot Hose