LMI Industrial Wall -Mount Controllers

Pre-wired electrical plugs permit easy connections
  • Pulse-output modules feature three point proportinal control
  • Use with Flow and level switches
Controllers with pulse output are compatible with 74501-, 07100-, and 74500-series pumps--no need for a separate timer to "translate" pulse signal.
440,000.00 - ₹725,000.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
LMI DP5000-1A-0 DP5000 Wall-Mount On/Off (Limit) pH Controller; 115 VAC 5730000 Mfr # DP5000-1A-0
COMPARE
DP5000 Wall-Mount On/Off (Limit) pH Controller; 115 VAC
₹516,000.00INR / Each
LMI DP5000-1A-1 pH on/off controller with isolated 4 to 20 mA output, 115 VAC 5730010 Mfr # W2T461739
COMPARE
pH on/off controller with isolated 4 to 20 mA output, 115 VAC
₹725,000.00INR / Each
LMI DP5000-1B-0 pH pulse output controller 115/230 VAC 5730020 Mfr # W2T461741
COMPARE
pH pulse output controller 115/230 VAC
₹440,000.00INR / Each
LMI DP5000-1B-1 pH pulse output controller with 4 to 20 mA output 115/230 VAC 5730030 Mfr # W2T461791
COMPARE
pH pulse output controller with 4 to 20 mA output 115/230 VAC
₹547,000.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat