Liquid Dispensing Carboy
Molded-in handles makes transport easy
  • Can be frozed, boiled, or sterlized
  • Easily dispense with spigot
These medium-density polyethylene liquid dispensers can be frozen, boiled, or sterilized. They have molded-in handles and a heavy-duty, no-drip faucet.
8,700.00 - ₹11,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Gal) Material Outside Diameter (OD) (in)
Availability Pricing
COMPARE
3 Polyethylene(PE) 10
₹8,700.00INR / Each
COMPARE
6 Polyethylene(PE) 12 1/4
₹11,000.00INR / Each