This product is now discontinued, View Related Products
Labconco XVS Ventilation Stations
Low air-flow technology for motion-sensitive operations
These sophisticated enclosures for balance usage and other low air-flow operations combine five air-flow components to produce a safe working environment without compromising the integrity of your manipulations. Volatile substances are handled without air-borne exposure to powders, sublimates, and fumes. Cabinet design also minimizes electrostatic effects and vibration to enable precise weighing results while eliminating the need for repeated enclosure clean-ups. Exhaust air to outside or return air to room through FilterMate Portable Exhauster.
Durable enclosures are constructed of epoxy-coated steel and aluminum for maximum resistance to corrosion and abrasion. Tempered safety glass sides, top, and front sash offer excellent visibility, ambient light, and protection. Glass is preferred over acrylic for optical clarity and resistance to solvent-induced hazing and marring. Ergonomic 20° angled sash operates glare-free with an 8” opening and pivots up to lock in position for 20”-clearance loading. Design and construction create unified resistance to vibration, electrostatic charges, and fire. Two utility ports for cords, cables, and tubing.
Select 22” or 31-1/4” enclosure height. All models require a remote blower or FilterMate Portable Exhauster, a work surface (See Accessories), and a supporting base.
XVS Ventilation Stations provide the same operating environment as XPert Balance Enclosures. Recommended for applications where airflow velocity is assured, e.g. with a nearby dedicated FilterMate unit or for non-critical outside exhaust applications.
579,000.00 - ₹817,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Exterior Depth (in) Exterior Height (in) Exterior Width (in)
Availability Pricing
Labconco 4862010 2 ft XVS Ventilation Station, 22" working height WW-09100-30 Mfr # 4862010
COMPARE
28.875 22.75 24
₹579,000.00INR / Each
Labconco 4862030 2 ft XVS Ventilation Station, 31-3/4" working height WW-09100-32 Mfr # 4862030
COMPARE
28.875 32 24
₹817,000.00INR / Each
Labconco 4863010 3 ft XVS Ventilation Station, 22" working height WW-09100-40 Mfr # 4863010
COMPARE
28.875 22.75 36
₹789,000.00INR / Each
Labconco 4863030 3 ft XVS Ventilation Station, 31-3/4" working height WW-09100-42 Mfr # 4863030
COMPARE
28.875 32 36
Call For Price
Labconco 4864010 4 ft XVS Ventilation Station, 22" working height WW-09100-50 Mfr # 4864010
COMPARE
28.875 22.75 48
Call For Price
Labconco 4864030 4 ft XVS Ventilation Station, 31-3/4" working height WW-09100-52 Mfr # 4864030
COMPARE
28.875 32 48