Labconco RXPert™ Class I Double-Filtered Biological Safety Cabinets

Keep hazardous powders and particulates contained
  • Meet USP <800> requirements for nonsterile hazardous compounding with redundant filtration optional
  • Bag in/bag out HEPA filter provides protection during filter changes
  • Outside ducting optional
The RXPert cabinet features a space-saving design constructed of a static-dissipative powder-coated aluminum frame, steel rear plenum and baffle, and tempered safety glass. The true bag-in/bag-out HEPA filter disposal system allows for safe filter replacement. The filtered balance system is a Class I enclosure that complies with USP requirements for nonsterile compounding.


All models include a built-in analog airflow monitor. Some models include a stainless steel or epoxy work surface; a work surface is required if one is not included.


Double-Filtered Enclosures feature redundant filters in series, so do not require exhausting to the outside while maintaining USP <800> compliance. All models require a supporting base (order separately).


Compliance: USP <800>, SEFA 1-2010, modified ASHRAE 110-1995, ANSI Z9.5-2011, UL 61010-1, and CAN/CSA C22.2 No. 61010.1 standards.
Call For Price

18 variations of this product are available.

See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Labconco RXPert  Double Filtered Balance System, Stainless Steel Work Surface, 2', 115 VAC 3252133 Mfr # 3971281
COMPARE
Double Filtered Balance System, Stainless Steel Work Surface, 2', 115 VAC
Call For Price
Labconco RXPert  Double Filtered Balance System, Epoxy Surface, 2', 115 VAC 3252135 Mfr # 3971282
COMPARE
Double Filtered Balance System, Epoxy Surface, 2', 115 VAC
Call For Price
Labconco RXPert  Double Filtered Balance System, Stainless Steel Work Surface, 3', 115 VAC 3252139 Mfr # 3971381
COMPARE
Double Filtered Balance System, Stainless Steel Work Surface, 3', 115 VAC
Call For Price
Labconco RXPert  Double Filtered Balance System, Epoxy Surface, 3', 115 VAC 3252141 Mfr # 3971382
COMPARE
Double Filtered Balance System, Epoxy Surface, 3', 115 VAC
Call For Price
Labconco RXPert  Double Filtered Balance System, Stainless Steel Work Surface, 4', 115 VAC 3252145 Mfr # 3971481
COMPARE
Double Filtered Balance System, Stainless Steel Work Surface, 4', 115 VAC
Call For Price
Labconco RXPert  Double Filtered Balance System, Epoxy Surface, 4', 115 VAC 3252147 Mfr # 3971482
COMPARE
Double Filtered Balance System, Epoxy Surface, 4', 115 VAC
Call For Price
Labconco RXPert  Tall Double Filtered Balance System, Stainless Steel Work Surface, 4', 115 V 3252151 Mfr # 3971486
COMPARE
Tall Double Filtered Balance System, Stainless Steel Work Surface, 4', 115 V
Call For Price
Labconco RXPert  Tall Double Filtered Balance System, Epoxy Surface, 4', 115 V 3252153 Mfr # 3971487
COMPARE
Tall Double Filtered Balance System, Epoxy Surface, 4', 115 V
Call For Price
Labconco RXPert  Tall & Deep Double Filtered Balance System, Stainless Steel, 4', 115 V 3252157 Mfr # 3971491
COMPARE
Tall & Deep Double Filtered Balance System, Stainless Steel, 4', 115 V
Call For Price
Labconco RXPert  Tall & Deep Double Filtered Balance System, Epoxy Surface, 4', 115 V 3252159 Mfr # 3971492
COMPARE
Tall & Deep Double Filtered Balance System, Epoxy Surface, 4', 115 V
Call For Price
Labconco RXPert  Tall Double Filtered Balance System, Stainless Steel Work Surface, 5', 115 V 3252163 Mfr # 3971586
COMPARE
Tall Double Filtered Balance System, Stainless Steel Work Surface, 5', 115 V
Call For Price
Labconco RXPert  Tall Double Filtered Balance System, Epoxy Surface, 5', 115 V 3252165 Mfr # 3971587
COMPARE
Tall Double Filtered Balance System, Epoxy Surface, 5', 115 V
Call For Price
Labconco RXPert  Tall & Deep Double Filtered Balance System, Stainless Steel, 5', 115 V 3252169 Mfr # 3971591
COMPARE
Tall & Deep Double Filtered Balance System, Stainless Steel, 5', 115 V
Call For Price
Labconco RXPert  Tall & Deep Double Filtered Balance System, Epoxy Surface, 5', 115 V 3252171 Mfr # 3971592
COMPARE
Tall & Deep Double Filtered Balance System, Epoxy Surface, 5', 115 V
Call For Price
Labconco RXPert  Tall Double Filtered Balance System, Stainless Steel Work Surface, 6', 115 V 3252175 Mfr # 3971686
COMPARE
Tall Double Filtered Balance System, Stainless Steel Work Surface, 6', 115 V
Call For Price
Labconco RXPert  Tall Double Filtered Balance System, Epoxy Surface, 6', 115 V 3252177 Mfr # 3971687
COMPARE
Tall Double Filtered Balance System, Epoxy Surface, 6', 115 V
Call For Price
Labconco RXPert  Tall & Deep Double Filtered Balance System, Stainless Steel, 6', 115 V 3252181 Mfr # 3971691
COMPARE
Tall & Deep Double Filtered Balance System, Stainless Steel, 6', 115 V
Call For Price
Labconco RXPert  Tall & Deep Double Filtered Balance System, Epoxy Surface, 6', 115 V 3252183 Mfr # 3971692
COMPARE
Tall & Deep Double Filtered Balance System, Epoxy Surface, 6', 115 V
Call For Price
Showing 1- 18 of 18

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat