Labconco RXPert Double Filtered Balance System, Epoxy Surface, 3', 115 VAC

Representative image only.

Labconco RXPert Double Filtered Balance System, Epoxy Surface, 3', 115 VAC

Labconco | Mfr: 3971382| Item: 3252141

Keep hazardous powders and particulates contained

Call For Price

Call For Price
Product Description

Includes

8-ft (2.4-m) power cord.

Key Features

  • Meet USP <800> requirements for nonsterile hazardous compounding with redundant filtration optional
  • Bag in/bag out HEPA filter provides protection during filter changes
  • Outside ducting optional
  • 1 year warranty

More About this Item

The RXPert cabinet features a space-saving design constructed of a static-dissipative powder-coated aluminum frame, steel rear plenum and baffle, and tempered safety glass. The true bag-in/bag-out HEPA filter disposal system allows for safe filter replacement. The filtered balance system is a Class I enclosure that complies with USP requirements for nonsterile compounding.


All models include a built-in analog airflow monitor. Some models include a stainless steel or epoxy work surface; a work surface is required if one is not included.


Double-Filtered Enclosures feature redundant filters in series, so do not require exhausting to the outside while maintaining USP <800> compliance. All models require a supporting base (order separately).


Compliance: USP <800>, SEFA 1-2010, modified ASHRAE 110-1995, ANSI Z9.5-2011, UL 61010-1, and CAN/CSA C22.2 No. 61010.1 standards.

1 Year Warranty
Specifications
  • Hood Width (in)36
  • Exterior Width (in)36
  • Exterior Height (in)49.5
  • Exterior Depth (in)29
  • Interior Width (in)33.625
  • Interior Height (in)22.75
  • Interior Depth (in)23.4375
  • Power (VAC)115
  • Power (Hz)60
  • Sash opening (in)9.4
  • Exhaust Volume (CFM)220
  • Shipping Weight (lbs)452
  • Shipping Weight (kg)205
  • DescriptionDouble Filtered Balance System, Epoxy Surface, 3', 115 VAC
  • Warranty1 Year
Reviews
Q&A