Labconco® Purifier® 6-Ft Wide Vertical Clean Benches
HEPA-filtered air directed downward protects your work from particulate contamination
  • Particulate-free ISO Class 5 air in work area
  • Supply HEPA filters offers 99.99% efficiency at 0.3 µm
  • Nominal downflow velocity of 45 to 65 ft/min
  • Differential pressure gauge helps determine HEPA filter loading
  • Lamp located above the work area to prevent glares
  • Ergonomically designed glass sash with a 10° slope provides more comfortable viewing
Purifier vertical clean benches with a 99.99% efficient HEPA filter provide ISO Class 5 conditions to protect your samples from particulate contamination. The room air is drawn downward through a prefilter to trap large particles, forced through a blower, filtered through a HEPA filter, then sent downward over the work area.
Variable speed blower with solid state control can be adjusted to extend filter life. Fluorescent lamp located above the work area and out of contact with the filtered air provides glare-free lighting. The blower and light switches are located conveniently within easy reach. The front-mounted differential pressure gauge helps determine when to replace the HEPA filter by monitoring the total system pressure across the HEPA filter and pressurized plenum.
The UV-opaque, UV-resistant, 1/4"-thick tempered safety glass sash with 10° slope offers maximum visibility, less glare, and more comfortable viewing to minimize operator fatigue. Easily pivot sash upward to open and lock for loading and cleaning. Pass tubing and electrical cords through the two utility ports with iris openings; seal ports with plugs (included) when not in use. Glacier white and gray powder-coated aluminum and steel construction provides excellent durability.
2,989,000.00 - ₹3,445,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Description
Availability Pricing
Labconco Purifier® Vertical Clean Bench, 6-Ft Wide; 115V WW-33517-96 Mfr # 3660600
COMPARE
115 6-Ft Wide Vertical Clean Bench; 115V
₹2,989,000.00INR / Each
Labconco Purifier® Vertical Clean Bench with UV Lamp, 6-Ft Wide; 115V WW-33517-97 Mfr # 3660601
COMPARE
115 6-Ft Wide Vertical Clean Bench with UV Lamp; 115V
₹3,197,000.00INR / Each
Labconco Purifier® Vertical Clean Bench with Airflow Monitor, 6-Ft Wide; 115V WW-33517-98 Mfr # 3660603
COMPARE
115 6-Ft Wide Vertical Clean Bench with Airflow Monitor; 115V
₹3,235,000.00INR / Each
Labconco Purifier® Vertical Clean Bench with UV Lamp & Airflow Monitor, 6-Ft Wide; 115V WW-33517-99 Mfr # 3660604
COMPARE
115 6-Ft Wide Vertical Clean Bench with UV Lamp & Airflow Monitor; 115V
₹3,445,000.00INR / Each
Labconco Purifier® Vertical Clean Bench, 6-Ft Wide; 230V WW-33518-48 Mfr # 3660620
COMPARE
230 6-Ft Wide Vertical Clean Bench; 230V
₹2,989,000.00INR / Each
Labconco Purifier® Vertical Clean Bench with UV Lamp, 6-Ft Wide; 230V WW-33518-49 Mfr # 3660621
COMPARE
230 6-Ft Wide Vertical Clean Bench with UV Lamp; 230V
₹3,197,000.00INR / Each
Labconco Purifier® Vertical Clean Bench with Airflow Monitor, 6-Ft Wide; 230V WW-33518-50 Mfr # 3660623
COMPARE
230 6-Ft Wide Vertical Clean Bench with Airflow Monitor; 230V
₹3,235,000.00INR / Each
Labconco Purifier® Vertical Clean Bench with UV Lamp & Airflow Monitor, 6-Ft Wide; 230V WW-33518-51 Mfr # 3660624
COMPARE
230 6-Ft Wide Vertical Clean Bench with UV Lamp & Airflow Monitor; 230V
₹3,445,000.00INR / Each
Showing 1- 8 of 8