Labconco Purifier® Vertical Clean Bench with UV Lamp, 6-Ft Wide; 115V

Representative image only.

Labconco Purifier® Vertical Clean Bench with UV Lamp, 6-Ft Wide; 115V

Labconco | Mfr: 3660601| Item: 3351797

HEPA-filtered air directed downward protects your work from particulate contamination

₹3,901,000.00INR / Each

₹3,901,000.00INR / Each

Buy It Together

Current Item

Labconco Purifier® Vertical Clean Bench with UV Lamp, 6-Ft Wide; 115V
Labconco Purifier® Vertical Clean Bench with UV Lamp, 6-Ft Wide; 115V
3351797

₹3,901,000.00INR / Each
+ =
Total Price 3914900.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Particulate-free ISO Class 5 air in work area
  • Supply HEPA filters offers 99.99% efficiency at 0.3 µm
  • Nominal downflow velocity of 45 to 65 ft/min
  • Differential pressure gauge helps determine HEPA filter loading
  • Lamp located above the work area to prevent glares
  • Ergonomically designed glass sash with a 10° slope provides more comfortable viewing
  • UV lamp provides secondary decontamination
  • 115 VAC operation
  • 1 year warranty

More About this Item

Purifier vertical clean benches with a 99.99% efficient HEPA filter provide ISO Class 5 conditions to protect your samples from particulate contamination. The room air is drawn downward through a prefilter to trap large particles, forced through a blower, filtered through a HEPA filter, then sent downward over the work area.

Variable speed blower with solid state control can be adjusted to extend filter life. Fluorescent lamp located above the work area and out of contact with the filtered air provides glare-free lighting. The blower and light switches are located conveniently within easy reach. The front-mounted differential pressure gauge helps determine when to replace the HEPA filter by monitoring the total system pressure across the HEPA filter and pressurized plenum.

The UV-opaque, UV-resistant, 1/4"-thick tempered safety glass sash with 10° slope offers maximum visibility, less glare, and more comfortable viewing to minimize operator fatigue. Easily pivot sash upward to open and lock for loading and cleaning. Pass tubing and electrical cords through the two utility ports with iris openings; seal ports with plugs (included) when not in use. Glacier white and gray powder-coated aluminum and steel construction provides excellent durability.

A 254 nm UV lamp offers added protection against contamination. Easily toggle between fluorescent light, UV light, and off using a 3-way switch. A protective coated-steel panel closes the sash opening to prevent inadvertent exposure to UV rays. Front-mounted control lets you select from eight time settings (5, 10, 15, 30, 60, 120, 240 minutes, or continuous) on the digital timer to provide timed UV light exposure.

The Purifier Vertical Clean Bench should never be used with biohazardous materials, toxins, acids, or radionuclides. Air from the work area is dispersed directly into the laboratory from the Clean Bench.

Canadian Standards AssociationUL Recognized1 Year Warranty
Specifications
  • LightsFluorescent
  • Exterior Width (in)72
  • Exterior Height (in)46
  • Exterior Depth (in)28.6
  • Interior Width (in)70.8
  • Interior Height (in)32
  • Interior Depth (in)25.3
  • Power (VAC)115
  • Power (Hz)50/60
  • Sash opening (in)11.6
  • Exhaust Volume (CFM)360 to 740
  • Filter TypePrefilter and HEPA
  • Description6-Ft Wide Vertical Clean Bench with UV Lamp; 115V
  • Warranty1 Year
Reviews
Q&A