Labconco Fume Hood Work Surfaces
  • Supports all Labconco benchtop fume hoods (work surface is required).
Work surfaces are solid epoxy resin and available flat or with SpillStopper surface, which have dished surfaces to keep accidental spills and liquids way from the seam between the hood wall and surrounding countertop. SpillStopper Surfaces support Protector Premier Hoods. Flat Surfaces support Fiberglass 30 and Basic 47 and 70 Hoods. They are offered without cutouts for a sink. Contact Cole-Parmer for work surfaces with pre-drilled cutouts.
242,000.00 - ₹767,000.00INR / Each
27 variations of this product are available.
See All Product Options Below
27 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hood Width (in) Description
Availability Pricing
Labconco SpillStopper Work Surface, Epoxy, 48" W x 30" D 0911531 Mfr # 9500400
COMPARE
48 SpillStopper Work Surface, epoxy, 48" W x 30" D
₹503,000.00INR / Each
Labconco SpillStopper Work Surface, Epoxy, 60" W x 30" D 0911533 Mfr # 9500500
COMPARE
60 SpillStopper Work Surface, epoxy, 60" W x 30" D
₹541,000.00INR / Each
Labconco SpillStopper Work Surface, Epoxy, 72" W x 30" D 0911535 Mfr # 9500600
COMPARE
72 SpillStopper Work Surface, epoxy, 72" W x 30" D
₹650,000.00INR / Each
Labconco Work Surface, flat epoxy, 36" 0911540 Mfr # 4882805
COMPARE
36 Work Surface, flat epoxy, 36"
₹273,000.00INR / Each
Labconco Work Surface, Flat Epoxy, 48" 0911542 Mfr # 4882803
COMPARE
36 Work surface, flat epoxy, 48"
Call For Price
Labconco Work Surface, Flat Epoxy, 72" 0911544 Mfr # 4882804
COMPARE
48 Work Surface, flat epoxy, 72"
₹552,000.00INR / Each
Labconco Work Surface, Flat Epoxy, 28" 0911546 Mfr # 4882806
COMPARE
28 Work Surface, flat epoxy, 28"
₹242,000.00INR / Each
Labconco Echo Filtered Fume Hood Work Surface 3', No Cupsink Cutouts 3253538 Mfr # 9501300
COMPARE
36 Filtered Fume Hood Work Surface 3', No Cupsink Cutouts
₹438,000.00INR / Each
Labconco Echo Filtered Fume Hood Work Surface 3', Right Rear Cupsink Cutouts 3253539 Mfr # 9501301
COMPARE
36 Filtered Fume Hood Work Surface 3', Right Rear Cupsink Cutouts
Call For Price
Labconco Echo Filtered Fume Hood Work Surface 3', Left Rear Cupsink Cutouts 3253540 Mfr # 9501310
COMPARE
36 Filtered Fume Hood Work Surface 3', Left Rear Cupsink Cutouts
Call For Price
Labconco Echo Filtered Fume Hood Work Surface 3', Left and Right Rear Cupsink Cutouts 3253541 Mfr # 9501311
COMPARE
36 Filtered Fume Hood Work Surface 3', Left and Right Rear Cupsink Cutouts
Call For Price
Labconco Echo Filtered Fume Hood Work Surface 4', No Cupsink Cutouts 3253542 Mfr # 9501400
COMPARE
48 Filtered Fume Hood Work Surface 4', No Cupsink Cutouts
₹548,000.00INR / Each
Labconco Echo Filtered Fume Hood Work Surface 4', Right Rear Cupsink Cutouts 3253543 Mfr # 9501401
COMPARE
48 Filtered Fume Hood Work Surface 4', Right Rear Cupsink Cutouts
Call For Price
Labconco Echo Filtered Fume Hood Work Surface 4', Left Rear Cupsink Cutouts 3253544 Mfr # 9501410
COMPARE
48 Filtered Fume Hood Work Surface 4', Left Rear Cupsink Cutouts
Call For Price
Labconco Echo Filtered Fume Hood Work Surface 4', Left and Right Rear Cupsink Cutouts 3253545 Mfr # 9501411
COMPARE
48 Filtered Fume Hood Work Surface 4', Left and Right Rear Cupsink Cutouts
Call For Price
Labconco Echo Filtered Fume Hood Work Surface 5', No Cupsink Cutouts 3253546 Mfr # 9501500
COMPARE
60 Filtered Fume Hood Work Surface 5', No Cupsink Cutouts
₹656,000.00INR / Each
Labconco Echo Filtered Fume Hood Work Surface 5', Right Rear Cupsink Cutouts 3253547 Mfr # 9501501
COMPARE
60 Filtered Fume Hood Work Surface 5', Right Rear Cupsink Cutouts
Call For Price
Labconco Echo Filtered Fume Hood Work Surface 5', Left Rear Cupsink Cutouts 3253548 Mfr # 9501510
COMPARE
60 Filtered Fume Hood Work Surface 5', Left Rear Cupsink Cutouts
Call For Price
Labconco Echo Filtered Fume Hood Work Surface 5', Left and Right Rear Cupsink Cutouts 3253549 Mfr # 9501511
COMPARE
60 Filtered Fume Hood Work Surface 5', Left and Right Rear Cupsink Cutouts
Call For Price
Labconco Echo Filtered Fume Hood Work Surface 6', No Cupsink Cutouts 3253550 Mfr # 9501600
COMPARE
72 Filtered Fume Hood Work Surface 6', No Cupsink Cutouts
₹767,000.00INR / Each
Showing 1 - 20 of 27