Labconco Echo Filtered Fume Hood Work Surface 6', No Cupsink Cutouts
Representative image only.
Labconco Echo Filtered Fume Hood Work Surface 6', No Cupsink Cutouts
Labconco | Mfr: 9501600| Item: 3253550
₹767,000.00INR / Each
₹767,000.00INR / Each
Buy It Together
Current Item
Labconco Echo Filtered Fume Hood Work Surface 6', No Cupsink Cutouts
Labconco Echo Filtered Fume Hood Work Surface 6', No Cupsink Cutouts
3253550
₹767,000.00INR / Each
+ =
Total Price 767000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Supports all Labconco benchtop fume hoods (work surface is required).
  • 1 year warranty
More About this Item
Work surfaces are solid epoxy resin and available flat or with SpillStopper surface, which have dished surfaces to keep accidental spills and liquids way from the seam between the hood wall and surrounding countertop. SpillStopper Surfaces support Protector Premier Hoods. Flat Surfaces support Fiberglass 30 and Basic 47 and 70 Hoods. They are offered without cutouts for a sink. Contact Cole-Parmer for work surfaces with pre-drilled cutouts.
ISO9001 Certified Supplier1 Year Warranty
Specifications
  • Hood Width (in)72
  • DescriptionFiltered Fume Hood Work Surface 6', No Cupsink Cutouts