Koflo Clear PVC Schedule 40 Pipe In-Line Static Mixers
Mix fluids as you pump them through a pipe line
  • Achieve a 100% mixed output
  • Visual check of your mixing process
Install these mixers directly into your process line to achieve a 100% mixed output—mixing elements are welded inside the pipe. All mixers have threaded ends. Choose PVC, 316 stainless steel, PVDF, or PFA. The clear PVC mixers are ideal for a visual check of your mixing processes. Models with removable elements are available on request—call our Application Specialists for details.
14,200.00 - ₹35,700.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mixer Length (in) Mounting Type No. Of Elements
Availability Pricing
Koflo 3/8-40C-4-6-2 Clear PVC schedule 40 pipe mixer; 3/8" NPT(M), 6 elements 0466985 Mfr # 3/8-40C-4-6-2
COMPARE
6.5 0.375" NPT(M) 6
₹14,200.00INR / Each
Koflo 3/8-40C-4-12-2 Clear PVC schedule 40 pipe mixer; 3/8" NPT(M), 12 elements 0466987 Mfr # 3/8-40C-4-12-2
COMPARE
11 0.375" NPT(M) 12
₹16,500.00INR / Each
Koflo 1/2-40C-4-6-2 Clear PVC schedule 40 pipe mixer; 1/2" NPT(M), 6 elements 0466989 Mfr # 1/2-40C-4-6-2
COMPARE
7 0.5" NPT(M) 6
₹18,500.00INR / Each
Koflo 1/2-40C-4-12-2 Clear PVC schedule 40 pipe mixer; 1/2" NPT(M), 12 elements 0466991 Mfr # 1/2-40C-4-12-2
COMPARE
12 0.5" NPT(M) 12
₹20,300.00INR / Each
Koflo 3/4-40C-4-6-2 Clear PVC schedule 40 pipe mixer; 3/4" NPT(M), 6 elements 0466992 Mfr # 3/4-40C-4-6-2
COMPARE
9 0.75" NPT(M) 6
₹19,600.00INR / Each
Koflo 3/4-40C-4-12-2 Clear PVC schedule 40 pipe mixer; 3/4" NPT(M), 12 elements 0466993 Mfr # 3/4-40C-4-12-2
COMPARE
15 0.75" NPT(M) 12
₹22,600.00INR / Each
Koflo 1-40C-4-6-2 Clear PVC schedule 40 pipe mixer; 1" NPT(M), 6 elements 0466994 Mfr # 1-40C-4-6-2
COMPARE
11 1" NPT(M) 6
₹21,700.00INR / Each
Koflo 1-40C-4-12-2 Clear PVC schedule 40 pipe mixer; 1" NPT(M), 12 elements 0466995 Mfr # 1-40C-4-12-2
COMPARE
18 1" NPT(M) 12
₹28,200.00INR / Each
Koflo 1-1/2-40C-4-6-2 Clear PVC schedule 40 pipe mixer; 1-1/2" NPT(M), 6 elements 0466996 Mfr # 1.5-40C-4-6-2
COMPARE
15 1.5" NPT(M) 6
₹26,100.00INR / Each
Koflo 1-1/2-40C-4-12-2 Clear PVC schedule 40 pipe mixer; 1-1/2" NPT(M), 12 elements 0466997 Mfr # 1.5-40C-4-12-2
COMPARE
28 1.5" NPT(M) 12
₹32,700.00INR / Each
Koflo 2-40C-4-6-2 Clear PVC schedule 40 pipe mixer; 2" NPT(M), 6 elements 0466998 Mfr # 2-40C-4-6-2
COMPARE
19 2" NPT(M) 6
₹31,500.00INR / Each
Koflo 2-40C-4-12-2 Clear PVC schedule 40 pipe mixer; 2" NPT(M), 12 elements 0466999 Mfr # 2-40C-4-12-2
COMPARE
35 2" NPT(M) 12
₹35,700.00INR / Each
Showing 1- 12 of 12