Cole-Parmer Septa for Vials
Choose the right septum for optimal performance and results
  • Various materials, thicknesses and hardnessess to suit your application
  • Pre-slit septa prevent a vacuum forming inside the vial
Red rubber/Polytetrafluoroethylene (PTFE): improved level of cleanliness compared to natural rubber. PTFE coating ensures compatibility.
Natural rubber/Ethylene Tetrafluoroethylene (TEF): features excellent resealing properties.
Silicone/PTFE: cleanest solution for contaminant free sample handling; autoclavable, too.
PTFE/Silicone/PTFE: PTFE laminated to both sides of high-purity silicone to prevent coring.
3,700.00 - ₹146,000.00INR / Pkg of 1000
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
Compareitem specifications
Outside Diameter (mm) Septum Type Special Features
Availability Pricing
COMPARE
8 PTFE 0.25 mm thick, 53° shore D
₹3,700.00INR / Pkg of 1000
COMPARE
8 Natural rubber/transparent TEF 1.0 mm thick, 60° shore A
₹12,800.00INR / Pkg of 1000
COMPARE
8 Red PTFE/white silicone/red PTFE 1.0 mm thick, 45° shore A
₹48,600.00INR / Pkg of 1000
COMPARE
8 White silicone/red PTFE 1.5 mm thick, 55° shore A
₹24,300.00INR / Pkg of 1000
COMPARE
8 Transparent blue silicone/white PTFE 1.3 mm thick, 45° shore A
₹39,800.00INR / Pkg of 1000
COMPARE
8 Transparent blue silicone/white PTFE with slit 1.3 mm thick, 45° shore A
₹35,600.00INR / Pkg of 1000
COMPARE
9 Red rubber/beige PTFE 1.0 mm thick, 45° shore A
₹22,600.00INR / Pkg of 1000
COMPARE
9 White silicone/red PTFE 1.5 mm thick, 55° shore A
₹33,500.00INR / Pkg of 1000
COMPARE
10 White silicone/red PTFE 1.3 mm thick, 45° shore A
₹37,900.00INR / Pkg of 1000
COMPARE
12 White silicone/blue PTFE with cross slit 1.5 mm thick, 55° shore A
₹78,800.00INR / Pkg of 1000
COMPARE
13 PTFE 0.25 mm thick, 53° shore D
₹4,500.00INR / Pkg of 1000
COMPARE
13 Natural rubber/transparent TEF 1.3 mm thick, 60° shore A
₹17,400.00INR / Pkg of 1000
COMPARE
13 Red PTFE/white silicone/red PTFE 1.0 mm thick, 45° shore A
₹65,700.00INR / Pkg of 1000
COMPARE
Natural PTFE/clear silicone 2.6 mm thick
₹7,100.00INR / Pkg of 100
COMPARE
16 White silicone/red PTFE 1.5 mm thick, 55° shore A
₹38,200.00INR / Pkg of 1000
COMPARE
16 Red butyl/gray PTFE 1.6 mm thick, 55° shore A
₹36,900.00INR / Pkg of 1000
COMPARE
18 White silicone/blue PTFE 1.7 mm thick, 45° shore A
₹53,600.00INR / Pkg of 1000
COMPARE
18 Transparent blue silicone (soft)/white PTFE 1.3 mm thick, 45° shore A
₹59,400.00INR / Pkg of 1000
COMPARE
18 Transparent blue silicone (soft)/white PTFE 3.2 mm thick, 45° shore A
₹75,100.00INR / Pkg of 1000
COMPARE
17.5 Transparent blue silicone/white PTFE 1.3 mm thick, 45° shore A
₹88,000.00INR / Pkg of 1000
Showing 1 - 20 of 28