Kinesis® Caps and Septa
Protect sample integrity by choosing the right accessories for analytical applications
  • Extend autosampler needle lifetime
  • Easy to use — no more over tightening/deforming septa
  • No more loss of samples with no sample evaporation
  • Prevent contamination and avoid damage to the needle and instrument by choosing the right septa for your application
Proper septa selection is critical to prevent contamination and avoid damage to the needle and instrument. Choose from a wide range of caps and septa that suit your vials and applications.
Caps are available in different materials (polypropylene, PTFE, silicone, polyethylene, and aluminum) and sizes from 8 mm to 11 mm in diameter. Septa for the caps are available slitted or non-slitted and bonded or non-bonded. Select color-coded caps and septa to quickly identify specific samples when handling multiple samples at the same time.
Range includes:
• Screw Caps
• Crimp Top Caps
• Snap Top Caps
Quality Standards:
The products are made from the highest quality materials and tested in accordance to the best industry standards.
More Details:
Choose from the widest range of Kinesis caps and septa for use in your chromatographic separations. The products are made from the highest quality materials and manufactured in ISO 9001 approved facility. All materials are produced in an ISO Class 8 (Class 100,000) cleanroom.
390.00 - ₹144,000.00INR / Case of 10000
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Description Cap Material
Availability Pricing
COMPARE
Kinesis Screw Caps, 9 mm, Blue PP, Non-Slitted/Non-Bonded, White PTFE/Red SL Septa; 100/Pk, 100 Pk/Cs Polypropylene
₹60,900.00INR / Case of 10000
COMPARE
Kinesis Screw Caps, 9 mm, Blue PP, Pre-Slit/Bonded, White PTFE/Red SL Septa; 100/Pk, 100 Pk/Cs Polypropylene
₹144,000.00INR / Case of 10000
COMPARE
Kinesis Screw Cap, 9 mm, Blue PP, Bonded, Clear PTFE/white Silicone Septa; 100/Pk
₹1,000.00INR / Pkg of 100
COMPARE
Kinesis Screw Caps, 9 mm, Blue PP, Non-Slitted/Non-Bonded, White PTFE/Red SL Septa; 100/Pk Polypropylene
₹390.00INR / Pkg of 100
COMPARE
Kinesis Screw Caps, 9 mm, Blue PP, Pre-Slit/Bonded, White PTFE/Red SL Septa; 100/Pk Polypropylene
₹1,500.00INR / Pkg of 100
COMPARE
Kinesis Screw Cap, 9 mm, Blue PP, Bonded, Clear PTFE/white Silicone Septa; 100/Pk, 100 Pk/Cs
Call For Price
COMPARE
Kinesis Crimp Top Caps, 11 mm, Aluminum, Red PTFE/White Silicone Septa; 100/Pk Aluminium
₹450.00INR / Pkg of 100
COMPARE
Kinesis Crimp Top Caps, 11 mm, Aluminum, Red PTFE/White Silicone Septa; 100/Pk, 100 Pk/Cs Aluminium
₹44,700.00INR / Case of 10000
COMPARE
Kinesis Snap-top PE Caps, 11 mm, White PTFE/red Silicone Septa, 100/Pk, 100 Pk/Cs Polyethylene
₹51,300.00INR / Case of 10000
COMPARE
Kinesis Snap-top PE Caps, 11 mm, White PTFE/red Silicone Septa, 100/Pk Polyethylene
₹510.00INR / Pkg of 100
COMPARE
Kinesis Snap-top PE Caps, 11 mm, Pre-Slit, White PTFE/Red Silicone Septa, 100/Pk, 100 Pk/Cs Polyethylene
₹55,900.00INR / Case of 10000
COMPARE
Kinesis Snap-top PE Caps, 11 mm, Pre-Slit, White PTFE/Red Silicone Septa, 100/Pk Polyethylene
₹560.00INR / Pkg of 100
COMPARE
Kinesis Snap-top PE Caps, 11 mm, Blue PTFE/White Silicone Septa, 100/Pk, 100 Pk/Cs Polyethylene
₹51,300.00INR / Case of 10000
COMPARE
Kinesis Snap-top PE Caps, 11 mm, Blue PTFE/White Silicone Septa, 100/Pk Polyethylene
₹510.00INR / Pkg of 100
COMPARE
Kinesis Snap-top PE Caps, 11 mm, Red PTFE/White Silicone Septa, 100/Pk, 100 Pk/Cs Polyethylene
₹51,300.00INR / Case of 10000
COMPARE
Kinesis Snap-top PE Caps, 11 mm, Red PTFE/White Silicone Septa, 100/Pk Polyethylene
₹510.00INR / Pkg of 100
COMPARE
Kinesis Screw Caps, 9 mm, Blue PP, Pre-Slit, Blue PTFE/White Silicone Septa; 100/Pk, 100 Pk/Cs Polypropylene
₹55,900.00INR / Case of 10000
COMPARE
Kinesis Screw Caps, 9 mm, Blue PP, Pre-Slit, Blue PTFE/White Silicone Septa; 100/Pk Polypropylene
₹560.00INR / Pkg of 100
Showing 1- 18 of 18