Cole-Parmer Snap-top PE Caps, 11 mm, White PTFE/red Silicone Septa, 100/Pk, 100 Pk/Cs
Representative image only.
Cole-Parmer Snap-top PE Caps, 11 mm, White PTFE/red Silicone Septa, 100/Pk, 100 Pk/Cs
Cole-Parmer | Item: 1634925
Protect sample integrity by choosing the right accessories for analytical applications
₹51,300.00INR / Case of 10000
₹51,300.00INR / Case of 10000
Product Description
Key Features
  • Extend autosampler needle lifetime
  • Easy to use — no more over tightening/deforming septa
  • No more loss of samples with no sample evaporation
  • Prevent contamination and avoid damage to the needle and instrument by choosing the right septa for your application
More About this Item
Proper septa selection is critical to prevent contamination and avoid damage to the needle and instrument. Choose from a wide range of caps and septa that suit your vials and applications.
Caps are available in different materials (polypropylene, PTFE, silicone, polyethylene, and aluminum) and sizes from 8 mm to 11 mm in diameter. Septa for the caps are available slitted or non-slitted and bonded or non-bonded. Select color-coded caps and septa to quickly identify specific samples when handling multiple samples at the same time.
Range includes:
• Screw Caps
• Crimp Top Caps
• Snap Top Caps
Quality Standards:
The products are made from the highest quality materials and tested in accordance to the best industry standards.
More Details:
Choose from the widest range of Kinesis caps and septa for use in your chromatographic separations. The products are made from the highest quality materials and manufactured in ISO 9001 approved facility. All materials are produced in an ISO Class 8 (Class 100,000) cleanroom.
Please Note: Vials, caps, and septa formerly under the Kinesis brand have now been rebranded Cole-Parmer. All the same great features and specifications, just with a new and improved look.
Specifications
  • Tube MaterialGlass
  • Cap TypeSnap-top
  • Cap MaterialPolyethylene
  • Cap colorTransparent
  • Septum TypeWhite PTFE/Red Silicone
  • SterileNo
  • DescriptionKinesis Snap-top PE Caps, 11 mm, White PTFE/red Silicone Septa, 100/Pk, 100 Pk/Cs