Kimtech Pure G3 Nitrile Cleanroom Gloves
Contain no latex proteins for sensitive workers
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
Kimtech Pure® cleanroom gloves are recommended for use in ISO Class 3 cleanroom or higher. These disposable gloves are made of a synthetic nitrile polymer and contain no natural rubber latex or silicone. Gloves feature textured fingertips for added grip, beaded cuffs for ease of donning, and a smooth finish to accommodate double donning. Each package includes sixty gloves, double-bagged in sealed cleanroom bags. Ambidextrous.
Compliance: ISO Class 3
13,300.00 - ₹17,400.00INR / Bag of 60
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size
Availability Pricing
Kimberly-Clark Kimtech Pure G3 NXT Nitrile Cleanroom Gloves, Size 7, 60/bag 8648531 Mfr # 11823
COMPARE
12 7
₹17,400.00INR / Bag of 60
Kimberly-Clark Kimtech Pure G3 NXT Nitrile Cleanroom Gloves, Size 7.5, 60/bag 8648532 Mfr # 11824
COMPARE
12 7.5
₹13,300.00INR / Bag of 60