Kimberly-Clark Kimtech Pure G3 NXT Nitrile Cleanroom Gloves, Size 7, 60/bag
Representative image only.
Kimberly-Clark Kimtech Pure G3 NXT Nitrile Cleanroom Gloves, Size 7, 60/bag
Kimberly-Clark | Mfr: 11823| Item: 8648531
Contain no latex proteins for sensitive workers
₹17,400.00INR / Bag of 60
₹17,400.00INR / Bag of 60
Product Description
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Kimtech Pure® cleanroom gloves are recommended for use in ISO Class 3 cleanroom or higher. These disposable gloves are made of a synthetic nitrile polymer and contain no natural rubber latex or silicone. Gloves feature textured fingertips for added grip, beaded cuffs for ease of donning, and a smooth finish to accommodate double donning. Each package includes sixty gloves, double-bagged in sealed cleanroom bags. Ambidextrous.
Compliance: ISO Class 3
Specifications
  • MaterialNitrile
  • Thickness (mil)4
  • Size7
  • Length (in)12
  • Length (cm)30.5
  • ColorWhite
  • GripTextured
  • CuffBeaded
  • ComplianceISO Class 3
  • SterileNo
  • DescriptionNitrile Cleanroom Gloves, Size 7, 60/bag