KIMCOTE Separatory Funnels
Ultraclear, plastic safety-coated glass funnels feature large lower stems that allow a column to "break" with the stopcock closed — allowing a more complete separation and eliminating the necessity of emptying the funnel. Feature glass stopper and autoclavable PTFE stopcock plug. Funnels meet ASTM specification E1096, Type 4 requirements.
197,000.00 - ₹487,000.00INR / Case of 4
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Stem Length (mm) Stopcock Plug Size
Availability Pricing
DWK Life Sciences (Kimble) KC29048T-125 Separatory Funnels With Glass Stopper, PTFE, 125 ml, 4/cs 0612502 Mfr # KC29048T-125
COMPARE
125 60 2
₹412,000.00INR / Case of 4
DWK Life Sciences (Kimble) KC29048T-250 Separatory Funnels With Glass Stopper, PTFE, 250 ml, 4/cs 0612504 Mfr # KC29048T-250
COMPARE
250 60 4
₹487,000.00INR / Case of 4
DWK Life Sciences (Kimble) KC29048T-1000 Separatory Funnel With Glass Stopper, PTFE, 1000 ml, 1/cs 0612508 Mfr # KC29048T-1000
COMPARE
1000 60 4
₹197,000.00INR / Case of 1