KimCote™ Safety-Coated Media Bottles
KimCote plastic coating for added safety
These ultra-clear, safety-coated Type 1 borosilicate glass media bottles feature wide opening for easy pouring. Bottles include permanent white enamel graduations and marking spots. Linerless blue polypropylene (PP) cap and clear PP pour ring are included. Both bottle and cap are autoclavable to 140°C (284°F).
9,600.00 - ₹37,000.00INR / Case of 4
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Max Graduation (mL)
Availability Pricing
DWK Life Sciences (Kimble) KC14395-100 Safety-Coated Media Bottle, Polypropylene Cap, 100 mL, 4/Cs 3452315 Mfr # KC14395-100
COMPARE
100 10 80
₹9,600.00INR / Case of 4
DWK Life Sciences (Kimble) KC14395-250 Safety-Coated Media Bottle, Polypropylene Cap, 250 mL, 4/Cs 3452317 Mfr # KC14395-250
COMPARE
250 25 200
₹10,500.00INR / Case of 4
DWK Life Sciences (Kimble) KC14395-500 Safety-Coated Media Bottle, Polypropylene Cap, 500 mL, 4/Cs 3452319 Mfr # KC14395-500
COMPARE
500 50 400
₹13,100.00INR / Case of 4
DWK Life Sciences (Kimble) KC14395-1000 Safety-Coated Media Bottle, Polypropylene Cap, 1 L, 4/Cs 3452321 Mfr # KC14395-1000
COMPARE
1000 50 900
₹18,200.00INR / Case of 4
DWK Life Sciences (Kimble) KC14395-2000 Safety-Coated Media Bottle, Polypropylene Cap, 2 L, 4/Cs 3452323 Mfr # KC14395-2000
COMPARE
2000 100 1800
₹27,200.00INR / Case of 4
DWK Life Sciences (Kimble) KC14395-5000 Safety-Coated Media Bottle, Polypropylene Cap, 5 L, 1/Cs 3452325 Mfr # KC14395-5000
COMPARE
5000 250 4000
₹19,100.00INR / Case of 1
DWK Life Sciences (Kimble) KC14395-10000 Safety-Coated Media Bottle, Polypropylene Cap, 10 L, 1/Cs 3452327 Mfr # KC14395-10000
COMPARE
10000 500 8000
₹37,000.00INR / Case of 1
Showing 1- 7 of 7