KIMBLE® FLEX-COLUMN® Chromatography Columns
Easy-to-use — column outlets permanently connected to barrel
  • Precision borosilicate glass columns provide inertness, visibility, and performance
  • Luer lock inlets and outlets on the columns provide simple and versatile tubing connections
  • Sterilize with 2N NaOH or 100% ethanol
KIMBLE® FLEX-COLUMN® chromatography columns are economical and easy-to-use. Short columns are typically used for ion exchange chromatography, sample desalting and semi-disposable applications, while the long columns are ideally suited for gel filtration and adsorption chromatography.
Columns are constructed of polypropylene reservoirs and column outlets permanently connected to a borosilicate glass barrel. Barrels are manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements. Bed support is manufactured from High Density Polyethylene (HDPE) with 20-micron nominal bed support porosity. Sterilize with 2N NaOH or 100% ethanol, autoclaving is not recommended.
9,100.00 - ₹85,900.00INR / Each
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Total Volume Capacity (mL) Column Inside Diameter (ID) (mm) Column Length (cm)
Availability Pricing
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 0.7 cm ID x 4 cm L, 2 mL; 1/Ea 3469300 Mfr # 420401-0704
COMPARE
2 7 4
₹9,100.00INR / Each
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 0.7 cm ID x 4 cm L, 2 mL; 10/CS 3469301 Mfr # 420400-0704
COMPARE
2 7 4
₹66,700.00INR / Case of 10
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 0.7 cm ID x 5 cm L, 2 mL; 10/CS 3469302 Mfr # 420400-0705
COMPARE
2 7 5
₹66,700.00INR / Case of 10
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 0.7 cm ID x 10 cm L, 4 mL; 1/Ea 3469303 Mfr # 420401-0710
COMPARE
4 7 10
₹11,300.00INR / Each
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 0.7 cm ID x 10 cm L, 4 mL; 10/CS 3469304 Mfr # 420400-0710
COMPARE
4 7 10
₹67,300.00INR / Case of 10
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 0.7 cm ID x 20 cm L, 8 mL; 1/Ea 3469305 Mfr # 420401-0720
COMPARE
8 7 20
₹11,300.00INR / Each
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 0.7 cm ID x 20 cm L, 8 mL; 10/CS 3469306 Mfr # 420400-0720
COMPARE
8 7 20
₹76,000.00INR / Case of 10
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 0.7 cm ID x 30 cm L, 12 mL; 1/Ea 3469307 Mfr # 420401-0730
COMPARE
12 7 30
₹13,900.00INR / Each
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 0.7 cm ID x 30 cm L, 12 mL; 10/CS 3469308 Mfr # 420400-0730
COMPARE
12 7 30
₹85,900.00INR / Case of 10
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 0.7 cm ID x 50 cm L, 20 mL; 1/Ea 3469309 Mfr # 420401-0750
COMPARE
20 7 50
₹13,900.00INR / Each
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 1.0 cm ID x 5 cm L, 4 mL; 1/Ea 3469310 Mfr # 420401-1005
COMPARE
4 10 5
₹9,200.00INR / Each
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 1.0 cm ID x 5 cm L, 4 mL; 10/CS 3469311 Mfr # 420400-1005
COMPARE
4 10 5
₹66,700.00INR / Case of 10
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 1.0 cm ID x 10 cm L, 8 mL; 1/Ea 3469312 Mfr # 420401-1010
COMPARE
8 10 10
₹11,400.00INR / Each
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 1.0 cm ID x 10 cm L, 8 mL; 10/CS 3469313 Mfr # 420400-1010
COMPARE
8 10 10
₹67,300.00INR / Case of 10
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 1.0 cm ID x 15 cm L, 12 mL; 1/Ea 3469314 Mfr # 420401-1015
COMPARE
12 10 15
₹11,400.00INR / Each
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 1.0 cm ID x 15 cm L, 12 mL; 10/CS 3469315 Mfr # 420400-1015
COMPARE
12 10 15
₹72,000.00INR / Case of 10
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 1.0 cm ID x 20 cm L, 16 mL; 1/Ea 3469316 Mfr # 420401-1020
COMPARE
16 10 20
₹11,400.00INR / Each
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 1.0 cm ID x 20 cm L, 16 mL; 10/CS 3469317 Mfr # 420400-1020
COMPARE
16 10 20
₹76,800.00INR / Case of 10
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 1.0 cm ID x 30 cm L, 24 mL; 1/Ea 3469318 Mfr # 420401-1030
COMPARE
24 10 30
₹11,400.00INR / Each
DWK Life Sciences (Kimble) FLEX-COLUMN® Chromatography Column, 1.0 cm ID x 30 cm L, 24 mL; 10/CS 3469319 Mfr # 420400-1030
COMPARE
24 10 30
₹76,800.00INR / Case of 10
Showing 1 - 20 of 50