Kimble-Chase Long-Tip Measuring (Mohr) Pipettes
Color-coded for ease in sorting and selecting the correct size pipet
  • Manufactured from borosilicate glass 5.1
  • Calibrated to deliver
  • Scale is permanent brown stain fused into uniform bore tubing without etching
  • Pipet is graduated to a base line which is on the straight tube above the taper
These pipettes are designed with small tip openings for chemical laboratory work.
40,200.00 - ₹46,300.00INR / Case of 12
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL)
Availability Pricing
DWK Life Sciences (Kimble) 37020-2 Long-Tip Measuring (Mohr) Pipettes, 2mL WW-25563-54 Mfr # 37020-2
COMPARE
2
₹40,200.00INR / Case of 12
DWK Life Sciences (Kimble) 37020-5 Long-Tip Measuring (Mohr) Pipettes, 5mL WW-25563-56 Mfr # 37020-5
COMPARE
5
₹40,200.00INR / Case of 12
DWK Life Sciences (Kimble) 37020-10 Long-Tip Measuring (Mohr) Pipettes, 10mL WW-25563-58 Mfr # 37020-10
COMPARE
10
₹46,300.00INR / Case of 12