DWK Life Sciences (Kimble) 37020-2 Long-Tip Measuring (Mohr) Pipettes, 2mL
Representative image only.
DWK Life Sciences (Kimble) 37020-2 Long-Tip Measuring (Mohr) Pipettes, 2mL
DWK Life Sciences (Kimble) | Mfr: 37020-2| Item: 2556354
Color-coded for ease in sorting and selecting the correct size pipet
₹52,400.00INR / Case of 12
₹52,400.00INR / Case of 12
Buy It Together
Current Item
DWK Life Sciences (Kimble) 37020-2 Long-Tip Measuring (Mohr) Pipettes, 2mL
DWK Life Sciences (Kimble) 37020-2 Long-Tip Measuring (Mohr) Pipettes, 2mL
2556354
₹52,400.00INR / Case of 12
+ =
Total Price 61700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Manufactured from borosilicate glass 5.1
  • Calibrated to deliver
  • Scale is permanent brown stain fused into uniform bore tubing without etching
  • Pipet is graduated to a base line which is on the straight tube above the taper
  • 1 year warranty
More About this Item
These pipettes are designed with small tip openings for chemical laboratory work.
1 Year Warranty
Specifications
  • SterileNo
  • Max Volume (µL)2000
  • Max Volume (mL)2
  • Max Dispensing (mL)2
  • DescriptionLong-TipMeasuring(Mohr)Pipettes,2mL