Kimberly-Clark G20 Atlantic Green Nitrile Gloves
Gloves offering Chemical splash protection
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Ambidextrous Green Nitrile Gloves
  • Chemical splash hand protection
  • Textured fingertips, beaded cuff
  • Food contact certified
Gloves providing chemical spalsh hand protection suitable for chemical handeling in non sterile environment and labs. Excellent tactile sensitivity.
Thickness at Palm: 2.4 mils thin
Thickness at fingertips: 3.5 mils thin
0.65 AQL for pinholes
25,300.00INR / Case
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Kimberly-Clark G20 Atlantic Green Nitrile Gloves Extra Small, 2500 Gloves/Cs WW-15964-95 Mfr # 90090
COMPARE
Atlantic Green Nitrile Gloves Extra Small, 2500 Gloves/Cs
Call For Price
Kimberly-Clark G20 Atlantic Green Nitrile Gloves Small, 2500 Gloves/Cs WW-15964-96 Mfr # 90091
COMPARE
Atlantic Green Nitrile Gloves Small, 2500 Gloves/Cs
₹25,300.00INR / Case
Kimberly-Clark G20 Atlantic Green Nitrile Gloves Medium, 2500 Gloves/Cs WW-15964-97 Mfr # 90092
COMPARE
Atlantic Green Nitrile Gloves Medium, 2500 Gloves/Cs
₹25,300.00INR / Case
Kimberly-Clark G20 Atlantic Green Nitrile Gloves Large, 2500 Gloves/Cs WW-15964-98 Mfr # 90093
COMPARE
Atlantic Green Nitrile Gloves Large, 2500 Gloves/Cs
₹25,300.00INR / Case
Kimberly-Clark G20 Atlantic Green Nitrile Gloves Extra Large, 2250 Gloves/Cs WW-15964-99 Mfr # 90094
COMPARE
Atlantic Green Nitrile Gloves Extra Large, 2250 Gloves/Cs