Kimberly-Clark G20 Atlantic Green Nitrile Gloves Large, 2500 Gloves/Cs
Representative image only.
Kimberly-Clark G20 Atlantic Green Nitrile Gloves Large, 2500 Gloves/Cs
Kimberly-Clark | Mfr: 90093| Item: 1596498
Gloves offering Chemical splash protection
₹50,800.00INR / Case
₹50,800.00INR / Case
Product Description
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Ambidextrous Green Nitrile Gloves
  • Chemical splash hand protection
  • Textured fingertips, beaded cuff
  • Food contact certified
More About this Item
Gloves providing chemical spalsh hand protection suitable for chemical handeling in non sterile environment and labs. Excellent tactile sensitivity.
Thickness at Palm: 2.4 mils thin
Thickness at fingertips: 3.5 mils thin
0.65 AQL for pinholes
Specifications
  • MaterialNitrile
  • SterileNo
  • DescriptionAtlantic Green Nitrile Gloves Large, 2500 Gloves/Cs