Kimax® BeakerPlus™ Combination Beaker and Flask
Don’t get splashed while mixing
  • Minimize splashing with narrow, recessed neck
  • Filter right into vessel— neck accommodates funnels and filters
Kimax® BeakerPlus™ Combination Beaker and Flask is ideal for mixing and for use with funnels or filters. The unique design features a wide fluted rim with pour spout and narrow recessed neck to reduce splashing during agitation. Manufactured with borosilicate glass, these beaker/flasks offer excellent chemical and heat resistance.
20,200.00 - ₹50,100.00INR / Pkg of 6
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
DWK Life Sciences (Kimble) 14035-150 BEAKERplus Combination Beaker and Flask, 150 mL, 6/pk WW-34517-00 Mfr # 14035-150
COMPARE
150 25 Borosilicate Glass
₹42,300.00INR / Pkg of 6
DWK Life Sciences (Kimble) 14035-250 BEAKERplus Combination Beaker and Flask, 250 mL, 6/pk WW-34517-02 Mfr # 14035-250
COMPARE
250 50 Borosilicate Glass
₹45,300.00INR / Pkg of 6
DWK Life Sciences (Kimble) 14035-500 BEAKERplus Combination Beaker and Flask, 500 mL, 6/pk WW-34517-04 Mfr # 14035-500
COMPARE
500 50 Borosilicate Glass
₹50,100.00INR / Pkg of 6
DWK Life Sciences (Kimble) 14035-1000 BEAKERplus Combination Beaker and Flask, 1000 mL, 2/pk WW-34517-06 Mfr # 14035-1000
COMPARE
1000 50 Borosilicate Glass
₹20,200.00INR / Pkg of 2
DWK Life Sciences (Kimble) 14035-1200 BEAKERplus Combination Beaker and Flask, 1200 mL, 2/pk WW-34517-08 Mfr # 14035-1200
COMPARE
1200 50 Borosilicate Glass
₹21,700.00INR / Pkg of 2