Kest Bottom Magnetic Mixer- RM Series
Outstanding mixing performance in critical pharma and biotechnology applications
  • Magnetic coupled mixer
  • Robust bearing design Zr/SiC. No particle shedding, under USP <788>
  • Designed for CIP/SIP applications
  • Easy operator handling of the drive unit using proprietary Kest-Lock
  • Advanced with aseptic design and excellent mixing performance
  • Speed Sensor technology (optional) for reliable process control
  • Mixing head designed to mix to the last drop
  • Engineering and customization options
  • Compliance with higher regulatory demands
Features:
• Designed to incorporate more energy into the tank (e.g. for dispersions)
• Mixing volume capacity: Up to 8KL
• Motor painted as per RAL 7015
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Kest Technology RM01 Drive Unit, Includes AC Motor and outer driving head; 1/Pk 1637987 Mfr # KM-RM-DU01-1
COMPARE
RM01 Drive Unit, Includes AC Motor and outer driving head; 1/Pk
Call For Price
Kest Technology RM03 Drive Unit, Includes AC Motor and outer driving head; 1/Pk 1637989 Mfr # KM-RM-DU03-1
COMPARE
RM03 Drive Unit, Includes AC Motor and outer driving head; 1/Pk
Call For Price
Kest Technology RM06 Drive Unit, Includes AC Motor and outer driving head; 1/Pk 1637991 Mfr # KM-RM-DU06-1
COMPARE
RM06 Drive Unit, Includes AC Motor and outer driving head; 1/Pk